Skip to Content
Merck
All Photos(1)

Key Documents

SML2997

Sigma-Aldrich

GHP-88309

≥98% (HPLC)

Synonym(s):

2-Fluoro-6-(5-isoquinolinyl)benzamide; 2-Fluoro-6-(5-isoquinolyl)benzamide, GHP 88309, GHP88309

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$189.00
25 MG
$761.00

$189.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$189.00
25 MG
$761.00

About This Item

Empirical Formula (Hill Notation):
C16H11FN2O
CAS Number:
Molecular Weight:
266.27
UNSPSC Code:
51111800
NACRES:
NA.77

$189.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC1=C(C(N)=O)C(C2=C(C=CN=C3)C3=CC=C2)=CC=C1

InChI key

OLNVLPGIADWVKU-UHFFFAOYSA-N

Biochem/physiol Actions

GHP-88309 is an orally available non-nucleoside paramyxovirus polymerase inhibitor with broad-spectrum anti-paramyxoviral activity (EC50 in μM= 0.4/HPIV3-JS, 0.8/HPIV3-5F6, 2/SeV-F1R-eGF, 0.6/MeV/Alaska.USA 16.00, 0.4/CDV-5804p) and little cytoxicity (CC50 >20μM). GHP-88309 potently inhibits the replication of HPIV1 isolate 5F6 and HPIV3 isolates 10L3 & 9R4 in 3D-ALI-HBTEC human airway organoids (EC50 = 280,90, 105 nM, respectively) in 3D-ALI-HBTECs. HPIV3 and provides complete protection against lethal Sendai virus infection in mice (150 mg/kg bid. po. starting at time of infection or 48 hrs after), a murine surrogate model of human HPIV3 disease.
Orally available non-nucleoside paramyxovirus polymerase inhibitor with broad-spectrum anti-paramyxoviral activity in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert M Cox et al.
Nature microbiology, 5(10), 1232-1246 (2020-07-15)
Paramyxoviruses such as human parainfluenza virus type-3 (HPIV3) and measles virus (MeV) are a substantial health threat. In a high-throughput screen for inhibitors of HPIV3 (a major cause of acute respiratory infection), we identified GHP-88309-a non-nucleoside inhibitor of viral polymerase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service