Skip to Content
Merck
All Photos(1)

Key Documents

SML2409

Sigma-Aldrich

NITD008

≥98% (HPLC)

Synonym(s):

7-(2-C-Ethynyl-b-D-ribofuranosyl)-7H-pyrrolo[2,3-d]pyrimidin-4-amine, 7-Deaza-2′-C-acetylene-adenosine, NITD 008

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$253.00
25 MG
$788.00

$253.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$253.00
25 MG
$788.00

About This Item

Empirical Formula (Hill Notation):
C13H14N4O4
CAS Number:
Molecular Weight:
290.27
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$253.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

NC1=C2C=CN(C2=NC=N1)[C@@H]3O[C@@H]([C@H]([C@](C#C)3O)O)CO

InChI

1S/C13H14N4O4/c1-2-13(20)9(19)8(5-18)21-12(13)17-4-3-7-10(14)15-6-16-11(7)17/h1,3-4,6,8-9,12,18-20H,5H2,(H2,14,15,16)/t8-,9-,12-,13-/m1/s1

InChI key

NKRAIOQPSBRMOV-NRMKKVEVSA-N

Biochem/physiol Actions

NITD008 is a selective inhibitor of flaviviruses including dengue virus (EC50 = 0.64 μM), hepatitis C virus and Zika virus, and has also been shown to inhibit tick-borne flaviviruses in vitro. NITD008 has been reported to inhibit RNA-dependent RNA polymerase (RdRp), an essential protein of RNA viruses.
Selective inhibitor of flaviviruses including dengue virus, hepatitis C virus, and Zika virus

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zheng Yin et al.
Proceedings of the National Academy of Sciences of the United States of America, 106(48), 20435-20439 (2009-11-18)
Dengue virus (DENV), a mosquito-borne flavivirus, is a major public health threat. The virus poses risk to 2.5 billion people worldwide and causes 50 to 100 million human infections each year. Neither a vaccine nor an antiviral therapy is currently
Michael K Lo et al.
Antiviral research, 130, 46-49 (2016-03-27)
There are currently no antiviral therapies available for the tick-borne flaviviruses associated with hemorrhagic fevers: Kyasanur Forest disease virus (KFDV), both classical and the Alkhurma hemorrhagic fever virus (AHFV) subtype, and Omsk hemorrhagic fever virus (OHFV). In this brief study
Amjad Ali et al.
Asian Pacific journal of tropical medicine, 10(4), 321-331 (2017-05-30)
Zika virus (ZIKV) is rapidly spreading across the America and its devastating outcomes for pregnant women and infants have driven this previously ignored pathogen into the limelight. Clinical manifestations are fever, joint pain or rash and conjunctivitis. Emergence of ZIKV

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service