Skip to Content
Merck
All Photos(1)

Key Documents

SML2978

Sigma-Aldrich

d9A-2

≥98% (HPLC)

Synonym(s):

N-(14-{[2-(2,6-dioxopiperidin-3-yl)-1,3-dioxo-2,3-dihydro-1H-isoindol-4-yl]amino}-3,6,9,12-tetraoxatetradecan-1-yl)-4′-fluoro-3′-methyl-2-[4-(4-methyl-1H-imidazol-5-yl)piperidin-1-yl]-[1,1′-biphenyl]-4-carboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$150.00
25 MG
$607.00

$150.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$150.00
25 MG
$607.00

About This Item

Empirical Formula (Hill Notation):
C46H54FN7O9
CAS Number:
Molecular Weight:
867.96
UNSPSC Code:
12352200
NACRES:
NA.77

$150.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(CCC1N2C(C(C=CC=C3NCCOCCOCCOCCOCCNC(C4=CC(N5CCC(C6=C(C)N=CN6)CC5)=C(C7=CC=C(F)C(C)=C7)C=C4)=O)=C3C2=O)=O)NC1=O

InChI key

WMBSSRAFHKGIRA-UHFFFAOYSA-N

Biochem/physiol Actions

d9A-2 is a potent degrader (PROTAC) of the solute carrier SLC9A1 impairs recovery from acid load and is potently cytotoxic to multiple cancer cell lines. Also, d9A-2 incites degradation of some SLC9 members including closely related SLC9A2 and SLC9A4, distant SLC9A7 and SLC9B1. d9A-2 comprise w9A ligand binding to SLC9A1 and the phthalimide-based CRBN binder.
potent degrader (PROTAC) of the solute carrier SLC9A1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ariel Bensimon et al.
Cell chemical biology, 27(6), 728-739 (2020-05-11)
With more than 450 members, the solute carrier (SLC) group of proteins represents the largest class of transporters encoded in the human genome. Their several-pass transmembrane domain structure and hydrophobicity contribute to the orphan status of many SLCs, devoid of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service