Skip to Content
Merck
All Photos(1)

Key Documents

5.05896

Sigma-Aldrich

Sumanirole Maleate

Synonym(s):

Sumanirole Maleate, Dopamine D₂ receptor Agonist, Sumanirole, PNU 95666, PNU95666

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$262.00

$262.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$262.00

About This Item

Empirical Formula (Hill Notation):
C11H13N3O · xC4H4O4
CAS Number:
Molecular Weight:
203.24 (free base basis)
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$262.00


Estimated to ship on29 May 2025


Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

solid

potency

9 nM Ki

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 16 mg/mL

storage temp.

−20°C

SMILES string

[n]21c3c([nH][c]2=O)cccc3C[C@H](C1)NC.OC(=O)\C=C/C(=O)O

InChI

1S/C11H13N3O.C4H4O4/c1-12-8-5-7-3-2-4-9-10(7)14(6-8)11(15)13-9;5-3(6)1-2-4(7)8/h2-4,8,12H,5-6H2,1H3,(H,13,15);1-2H,(H,5,6)(H,7,8)/b;2-1-/t8-;/m1./s1

InChI key

VOJRMYBBPKNLLI-ORHWHDKWSA-N

General description

An extremely selective dopamine D2 receptor agonist (Ki = 9 nM). Commonly used in Parkinson′s Disease models and to reproduce cocaine′s discriminative stimulus effects. Sumanirole has greater than 200-fold selectivity for the D2 receptor subtype versus the other dopamine receptor subtypes in radioligand binding assays. In cell-based assays, sumanirole is a fully efficacious agonist, with EC50 values between 17 and 75 nM. In animals, sumanirole elicits many physiological responses attributed to D2-like receptor function.

Warning

Toxicity: Standard Handling (A)

Other Notes

Achat-Mendes, C. et al., 2010. J. Pharmacol. Exp. Ther.334, 556.
McCall, R. B. et al., 2005. J. Pharmacol. Exp. Ther.314, 1248.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert B McCall et al.
The Journal of pharmacology and experimental therapeutics, 314(3), 1248-1256 (2005-06-28)
The purpose of this study is to demonstrate that sumanirole is a novel dopamine receptor agonist with high in vitro and in vivo selectivity for the D(2) receptor subtype. Sumanirole, (R)-5,6-dihydro-5-(methylamino)-4H-imidazo[4,5,1-ij]quinolin-2(1H)-one (Z)-2-butenedioate (1:1), is unique; it has greater than 200-fold
Cindy Achat-Mendes et al.
The Journal of pharmacology and experimental therapeutics, 334(2), 556-565 (2010-05-25)
Dopamine (DA) D3 and D2 receptor mechanisms are implicated in cocaine's abuse-related behavioral effects, but the relative contribution of the two receptor subtypes is only partially characterized. This study investigated the role of D3 and D2 subtype mechanisms by determining

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service