Skip to Content
Merck
All Photos(1)

Key Documents

SML2929

Sigma-Aldrich

Bilastine

≥98% (HPLC)

Synonym(s):

2-[4-(2-(4-(1-(2-ethoxyethyl)benzimidazole-2-yl)piperidine-1-yl)ethyl)phenyl]-2-methyl-propanoic acid, 4-[2-[4-[1-(2-Ethoxyethyl)-1H-benzimidazol-2-yl]-1-piperidinyl]ethyl]-α,α-dimethylbenzeneacetic acid, F-96221-BM1

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$146.00
25 MG
$593.00

$146.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$146.00
25 MG
$593.00

About This Item

Empirical Formula (Hill Notation):
C28H37N3O3
CAS Number:
Molecular Weight:
463.61
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$146.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[N+H]2(CCC(CC2)c3[n](c4c(n3)cccc4)CCOCC)CCc1ccc(cc1)C(C)(C)C(=O)[O-]

InChI

1S/C28H37N3O3/c1-4-34-20-19-31-25-8-6-5-7-24(25)29-26(31)22-14-17-30(18-15-22)16-13-21-9-11-23(12-10-21)28(2,3)27(32)33/h5-12,22H,4,13-20H2,1-3H3,(H,32,33)

InChI key

ACCMWZWAEFYUGZ-UHFFFAOYSA-N

Biochem/physiol Actions

Bilastine is an orally available, potent and selective histamine H1 receptor antagonist that exhibits long drug-target residence time at H1 receptor (73 min), which results in extended H1 receptor antagonism in vitro. Bilastine is used for treatment of allergic rhinitis and urticaria (hives).
orally available, potent and selective histamine H1 receptor antagonist that exhibits long drug-target residence time at H1 receptor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reyes Corcóstegui et al.
Drugs in R&D, 6(6), 371-384 (2005-11-09)
This study aimed to establish the receptor selectivity and antihistaminic activity of bilastine, a new selective antihistamine receptor antagonist. In vitro experiments were conducted using a receptor binding screening panel and guinea-pig and rat tissues. Antihistaminic activity was determined using
Xue Yan Wang et al.
Therapeutics and clinical risk management, 12, 585-597 (2016-04-26)
Allergic rhinitis and urticaria are common allergic diseases that may have a major negative impact on patients' quality of life. Bilastine, a novel new-generation antihistamine that is highly selective for the H1 histamine receptor, has a rapid onset and prolonged
Reggie Bosma et al.
European journal of pharmacology, 838, 107-111 (2018-09-12)
Drug-target binding kinetics has recently attracted considerable interest in view of the potential predictive power for in vivo drug efficacy. The recently introduced antihistamine bilastine has a long duration of in vivo drug action, which outlasts pharmacological active bilastine concentrations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service