Skip to Content
Merck
All Photos(1)

Key Documents

SML2891

Sigma-Aldrich

M871 trifluoroacetate

≥95% (HPLC)

Synonym(s):

Galanin-(2-13)-Glu-His-(Pro)3-(Ala-Leu)2-Ala-amide, TFA salt, M 871, TFA salt, M-871, TFA salt, Trp-Thr-Leu-Asn-Ser-Ala-Gly-Tyr-Leu-Leu-Gly-Pro-Glu-His-Pro-Pro-Pro-Ala-Leu-Alal-Leu-Ala-NH2, TFA salt, WTLNSAGYLLGPEHPPPALALA-NH2, TFA salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$257.00
5 MG
$852.00

$257.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$257.00
5 MG
$852.00

About This Item

Empirical Formula (Hill Notation):
C108H163N27O28 · xC2HF3O2
Molecular Weight:
2287.61 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

$257.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized powder

color

white to off-white

storage temp.

−20°C

Biochem/physiol Actions

M871 is a type 2-selective galanin receptor antagonist (GalR2/GalR1 Ki = 13.1/410 nM by competitive binding against 0.1 nM porcine galanin using membranes from CHO expressing respective human receptors; GalR2/GalR1 Ki = 2.5/1.1 nM in case of rat galanin) that potently blocks 10 nM rat galanin-induced inositol phosphate production in human GalR2-expressing CHO cells (by 100% with 0.1, 1 or 10 nM M871). M871 is widely employed in cultures (0.1 nM-2.5 μM) and in mice and rats in vivo via ip., brain region- or other site-specific injections.
Selective galanin receptor type 2 (GalR2) antagonist with in vitro and in vivo efficacy.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jesus-Servando Medel-Matus et al.
Experimental neurology, 289, 64-72 (2016-12-26)
Status epilepticus (SE) in rats, along with chronic epilepsy, leads to the development of behavioral impairments resembling depressive disorder and/or attention deficit/hyperactivity disorder (ADHD), thus reflecting respective comorbidities in epilepsy patients. Suppressed neurotransmitter tone in the raphe nucleus (RN)-prefrontal cortex
Seu-Hwa Chen et al.
PloS one, 13(6), e0199512-e0199512 (2018-06-22)
In this study, we investigated temporal changes in galanin receptor type 2 (GalR2) expression in NF200-, galanin-, neuropeptide Y (NPY)-, and neuronal nitric oxide synthase (nNOS)-like immunoreactive (LI) dorsal root ganglion (DRG) neurons after median nerve chronic constriction injury (CCI)
Pawel A Kolodziejski et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1863(10), 1228-1236 (2018-10-12)
Spexin (SPX, NPQ) is a novel peptide involved in the regulation of energy metabolism. SPX inhibits food intake and reduces body weight. In obese humans, SPX is the most down-regulated gene in fat. Therefore, SPX might be involved in the
M871-A Novel Peptide Antagonist Selectively Recognizing the Galanin Receptor Type 2
Sollenberg UE, Lundstrom L, Bartfai T, Langel U
International Journal of Peptide Research and Therapeutics, 12, 115-119 (2006)
Penghua Fang et al.
Biochemical pharmacology, 156, 241-247 (2018-09-01)
Although recent results of our and other studies have showed that galanin (GAL) is an antidiabetic and anti-inflammatory neuropeptide, the molecular mechanism how central GAL regulates energy homeostasis and insulin sensitivity is still not fully understood. The aim of this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service