Skip to Content
Merck
All Photos(1)

Key Documents

567726

Sigma-Aldrich

D-erythro-Sphingosine, Free Base, High Purity

Highly purified preparation of bovine brain sphingosine containing >99% of the erythro isomer.

Synonym(s):

D-erythro-Sphingosine, Free Base, High Purity, Cerebroside, trans-D- erythro-2-Amino-4-octadecene-1,3-diol, Ceramide, Cerebroside, trans-D-erythro-2-Amino-4-octadecene-1,3-diol, Ceramide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$281.00

$281.00


Estimated to ship on27 May 2025



Select a Size

Change View
10 MG
$281.00

About This Item

Empirical Formula (Hill Notation):
C18H37NO2
CAS Number:
Molecular Weight:
299.49
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77

$281.00


Estimated to ship on27 May 2025


Quality Level

description

Merck USA index - 14, 8747

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

chloroform: 1 mg/mL
methanol: 1 mg/mL
DMSO: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

N[C@H]([C@H](O)\C=C\CCCCCCCCCCCCC)CO

InChI

1S/C18H37NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h14-15,17-18,20-21H,2-13,16,19H2,1H3/b15-14+/t17-,18+/m0/s1

InChI key

WWUZIQQURGPMPG-KRWOKUGFSA-N

General description

Highly purified preparation of bovine brain sphingosine containing >99% of the erythro isomer. A potent, cell-permeable, reversible, and selective inhibitor of protein kinase C (PKC; IC50 = 2.8 µM) and insulin receptor tyrosine kinase. PKC inhibition is competitive with respect to diacylglycerol, phorbol dibutyrate, and Ca2+. Does not affect the activity of myosin light chain kinase and protein kinase A. Induces apoptosis in human leukemia HL-60 cells. Note: This product is not to be used for animal treatment, in vivo research or in any other contact procedure with livestock.

Biochem/physiol Actions

Cell permeable: no
Primary Target
PKC
Product does not compete with ATP.
Reversible: no
Target IC50: 2.8 µM against PKC

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot, purge with N₂ gas, and freeze (-20°C). Stock solutions are stable for up to 6 months under inert atmosphere at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service