Skip to Content
Merck
All Photos(1)

Key Documents

SML2752

Sigma-Aldrich

ZINC37781618

≥98% (HPLC)

Synonym(s):

2-Chloro-N-[3-(3-methyl-1H-1,2,4-triazol-5-yl)phenyl]-benzenemethanamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$114.00
25 MG
$463.00

$114.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$114.00
25 MG
$463.00

About This Item

Empirical Formula (Hill Notation):
C16H15ClN4
CAS Number:
Molecular Weight:
298.77
UNSPSC Code:
12352200
NACRES:
NA.77

$114.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Clc1c(cccc1)CNc2cc(ccc2)c3[nH]c(nn3)C

InChI

1S/C16H15ClN4/c1-11-19-16(21-20-11)12-6-4-7-14(9-12)18-10-13-5-2-3-8-15(13)17/h2-9,18H,10H2,1H3,(H,19,20,21)

InChI key

BNIQNBVKGOYHPC-UHFFFAOYSA-N

Biochem/physiol Actions

ZINC37781618 is an inactive control for selective melatonin MT1 receptor inverse agonist UCSF7447 (ZINC555417447).
inactive control for selective melatonin MT1 receptor inverse agonist UCSF7447

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reed M Stein et al.
Nature, 579(7800), 609-614 (2020-02-11)
The neuromodulator melatonin synchronizes circadian rhythms and related physiological functions through the actions of two G-protein-coupled receptors: MT1 and MT2. Circadian release of melatonin at night from the pineal gland activates melatonin receptors in the suprachiasmatic nucleus of the hypothalamus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service