Skip to Content
Merck
All Photos(1)

Key Documents

SML2751

Sigma-Aldrich

UCSF7447

≥98% (HPLC)

Synonym(s):

2-Bromo-4-chloro-N-[3-(1H-1,2,4-triazol-5-yl)phenyl]-benzenemethanamine, ZINC555417447

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$545.00

$124.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$124.00
25 MG
$545.00

About This Item

Empirical Formula (Hill Notation):
C15H12BrClN4
CAS Number:
Molecular Weight:
363.64
UNSPSC Code:
12352200
NACRES:
NA.77

$124.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

Brc1c(ccc(c1)Cl)CNc2cc(ccc2)c3[nH]ncn3

InChI

1S/C15H12BrClN4/c16-14-7-12(17)5-4-11(14)8-18-13-3-1-2-10(6-13)15-19-9-20-21-15/h1-7,9,18H,8H2,(H,19,20,21)

InChI key

QPTCYEYTAWEZNT-UHFFFAOYSA-N

Biochem/physiol Actions

UCSF7447 (ZINC555417447) brain permeable, potent and selective inverse agonist of both human and mouse melatonin MT1 receptor, which decelerates re-entrainment in mouse model of east-bound jetlag. Unexpectedly UCSF7447 advances the phase of the mouse circadian clock when administer at dusk.
brain permeable, potent and selective inverse agonist of both human and mouse melatonin MT1 receptor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reed M Stein et al.
Nature, 579(7800), 609-614 (2020-02-11)
The neuromodulator melatonin synchronizes circadian rhythms and related physiological functions through the actions of two G-protein-coupled receptors: MT1 and MT2. Circadian release of melatonin at night from the pineal gland activates melatonin receptors in the suprachiasmatic nucleus of the hypothalamus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service