Skip to Content
Merck
All Photos(1)

Key Documents

SML2737

Sigma-Aldrich

AER-271

≥98% (HPLC)

Synonym(s):

2-(3,5-bis(Trifluoromethyl)phenylcarbamoyl)-4-chlorophenyl dihydrogen phosphate, AER-270 phosphate, AER-270 precursor, AER-270 prodrug, IMD-0354 phosphate, IMD-0354 precursor, IMD-0354 prodrug, N-[3,5-bis(Trifluoromethyl)phenyl]-5-chloro-2-(phosphonooxy)benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$163.00
25 MG
$528.00

$163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$163.00
25 MG
$528.00

About This Item

Empirical Formula (Hill Notation):
C15H9ClF6NO5P
CAS Number:
Molecular Weight:
463.65
UNSPSC Code:
12352200
NACRES:
NA.77

$163.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)c1cc(cc(c1)C(F)(F)F)NC(=O)c2c(ccc(c2)Cl)O[P](=O)(O)O

InChI key

WSHXPHFIHYXZKC-UHFFFAOYSA-N

Biochem/physiol Actions

AER-271 is a phosphonate precursor (prodrug) form of the selective aquaporin 4 (AQP4) inhibitor IMD-0354 (AER-270; hu/m/r IC50 = 420/390/210 nM using respective CHO AQP4 transfectants) with >5000-fold enhanced solubility. AER-271 is rapidly converted to AER-270 in vivo (plasma AER-270 Cmax = 500 ng/mL or 1.75 μM, 20 min post 10 mg AER-271/kg i.p.; C57BL/6 mice) and exhibits therapeutic efficacy in rodent models of brain damage by cardiac arrest or ischemic stroke (5-10 mg/kg ip. & 0.08 mg/h sc. in mice; 1-10 mg/kg iv. in rats).
Phosphonate precursor (prodrug) of the selective aquaporin 4 (AQP4) inhibitor IMD-0354 (AER-270) with in vivo efficacy in rodent models of brain damage.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Nicosia et al.
Scientific reports, 9(1), 7417-7417 (2019-05-17)
Aquaporins (AQPs) are water channels that mediate a variety of biological processes. However, their role in the immune system is poorly understood. We recently reported that AQP4 is expressed by naïve and memory T cells and that AQP4 blockade with
Katayoun Ayasoufi et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 18(5), 1238-1246 (2017-12-16)
Prolonged cold ischemia storage (CIS) is a leading risk factor for poor transplant outcome. Existing strategies strive to minimize ischemia-reperfusion injury in transplanted organs, yet there is a need for novel approaches to improve outcomes of marginal allografts and expand
George W Farr et al.
Neuroscience, 404, 484-498 (2019-02-10)
Cerebral edema in ischemic stroke can lead to increased intracranial pressure, reduced cerebral blood flow and neuronal death. Unfortunately, current therapies for cerebral edema are either ineffective or highly invasive. During the development of cytotoxic and subsequent ionic cerebral edema
Ankit Awasthi et al.
Science (New York, N.Y.), 363(6422) (2018-12-14)
Forgetting is important. Without it, the relative importance of acquired memories in a changing environment is lost. We discovered that synaptotagmin-3 (Syt3) localizes to postsynaptic endocytic zones and removes AMPA receptors from synaptic plasma membranes in response to stimulation. AMPA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service