Merck
  • Synaptotagmin-3 drives AMPA receptor endocytosis, depression of synapse strength, and forgetting.
Synaptotagmin-3 drives AMPA receptor endocytosis, depression of synapse strength, and forgetting.
Science (New York, N.Y.) (2018-12-14)
Ankit Awasthi, Binu Ramachandran, Saheeb Ahmed, Eva Benito, Yo Shinoda, Noam Nitzan, Alina Heukamp, Sabine Rannio, Henrik Martens, Jonas Barth, Katja Burk, Yu Tian Wang, Andre Fischer, Camin Dean
ABSTRACT
Forgetting is important. Without it, the relative importance of acquired memories in a changing environment is lost. We discovered that synaptotagmin-3 (Syt3) localizes to postsynaptic endocytic zones and removes AMPA receptors from synaptic plasma membranes in response to stimulation. AMPA receptor internalization, long-term depression (LTD), and decay of long-term potentiation (LTP) of synaptic strength required calcium-sensing by Syt3 and were abolished through Syt3 knockout. In spatial memory tasks, mice in which Syt3 was knocked out learned normally but exhibited a lack of forgetting. Disrupting Syt3:GluA2 binding in a wild-type background mimicked the lack of LTP decay and lack of forgetting, and these effects were occluded in the Syt3 knockout background. Our findings provide evidence for a molecular mechanism in which Syt3 internalizes AMPA receptors to depress synaptic strength and promote forgetting.
Sigma-Aldrich
Anti-NMDAR2A Antibody, Chemicon®, from mouse
Sigma-Aldrich
F13714, ≥95% (HPLC)
Sigma-Aldrich
Anti-Glutamate Receptor 2 Antibody, extracellular, clone 6C4, clone 6C4, Chemicon®, from mouse
Sigma-Aldrich
AER-271, ≥98% (HPLC)
Sigma-Aldrich
Anti-NMDAR1 Antibody, rabbit monoclonal, culture supernatant, clone 1.17.2.6, Chemicon®
Sigma-Aldrich
AMG517, ≥98% (HPLC)