Skip to Content
Merck
All Photos(1)

Key Documents

SML1797

Sigma-Aldrich

L67

≥98% (HPLC)

Synonym(s):

LigI/III inhibitor L67, N-(3,5-Dibromo-4-methylphenyl)-glycine 2-[(2-hydroxy-5-nitrophenyl)methylene]hydrazide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$257.00

$257.00


Available to ship on11 April 2025Details



Select a Size

Change View
5 MG
$257.00

About This Item

Empirical Formula (Hill Notation):
C16H14Br2N4O4
CAS Number:
Molecular Weight:
486.11
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$257.00


Available to ship on11 April 2025Details


Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(N/N=C/C1=CC([N+]([O-])=O)=CC=C1O)CNC2=CC(Br)=C(C)C(Br)=C2

InChI key

KFEPVFJQVOMODD-IFRROFPPSA-N

Biochem/physiol Actions

L67 is a potent and specific inhibitor of DNA ligase IIIα (LigIIIα) that preferentially targets mitochondrial LigIIIα resulting in mitochondrial dysfunction.
L67 is a potent and specific inhibitor of DNA ligase IIIα (LigIIIα) that preferentially targets mitochondrial LigIIIα resulting in mitochondrial dysfunction. L67 preferentially targets cancer cell mitochondria resulting in enhanced ROS production and caspase 1-dependent apoptosis. L67 in combination with PARP inhibitors decreases survival rate of therapy resistant breast cancer and leukemia cells.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xi Chen et al.
Cancer research, 68(9), 3169-3177 (2008-05-03)
Based on the crystal structure of human DNA ligase I complexed with nicked DNA, computer-aided drug design was used to identify compounds in a database of 1.5 million commercially available low molecular weight chemicals that were predicted to bind to
L A Tobin et al.
Oncogene, 32(14), 1784-1793 (2012-05-30)
Resistance to imatinib (IM) and other tyrosine kinase inhibitors (TKI)s is an increasing problem in leukemias caused by expression of BCR-ABL1. As chronic myeloid leukemia (CML) cell lines expressing BCR-ABL1 utilize an alternative non-homologous end-joining pathway (ALT NHEJ) to repair
Annahita Sallmyr et al.
Cancer research, 76(18), 5431-5441 (2016-08-10)
Elevated levels of DNA ligase IIIα (LigIIIα) have been identified as a biomarker of an alteration in DNA repair in cancer cells that confers hypersensitivity to a LigIIIα inhibitor, L67, in combination with a poly (ADP-ribose) polymerase inhibitor. Because LigIIIα
Rajeswari Jayavaradhan et al.
Journal of molecular biology, 431(1), 102-110 (2018-05-12)
The efficient site-specific DNA double-strand breaks (DSB) created by CRISPR/Cas9 has revolutionized genome engineering and has great potential for editing hematopoietic stem/progenitor cells (HSPCs). However, detailed understanding of the variables that influence choice of DNA-DSB repair (DDR) pathways by HSPC

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service