Skip to Content
Merck
All Photos(1)

Key Documents

SML1784

Sigma-Aldrich

BTI-A-404

≥98% (HPLC)

Synonym(s):

4-[4-(Dimethylamino)phenyl]-N-(3,5-dimethylphenyl)-1,2,3,4-tetrahydro-6-methyl-2-oxo-5-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$203.00

$203.00


Check Cart for Availability


Select a Size

Change View
5 MG
$203.00

About This Item

Empirical Formula (Hill Notation):
C22H26N4O2
CAS Number:
Molecular Weight:
378.47
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

$203.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1NC(C2=CC=C(N(C)C)C=C2)C(C(NC3=CC(C)=CC(C)=C3)=O)=C(C)N1

InChI key

JQVNMIXFWKEYQJ-UHFFFAOYSA-N

Biochem/physiol Actions

BTI-A-404 is a potent and selective competitive inverse agonist of human GPR43 (FFAR2).
BTI-A-404 is a potent and selective competitive inverse agonist of human GPR43 (GPR43/Free Fatty Acid Receptor 2, FFAR2) that increases the cyclic AMP level and regulates and reduces acetate-induced cytoplasmic Ca2+ level. BTI-A-404 regulates the downstream signaling pathways of GPR43, including ERK, p38 MAPK, and NF-KB. BTI-A-404 stimulates GLP-1 release.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service