Skip to Content
Merck
All Photos(1)

Key Documents

SML1782

Sigma-Aldrich

CATPB

≥98% (HPLC)

Synonym(s):

(S)-3-(2-(3-Chlorophenyl)acetamido)-4-(4-(trifluoromethyl)phenyl)butanoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$204.00

$204.00


Check Cart for Availability


Select a Size

Change View
5 MG
$204.00

About This Item

Empirical Formula (Hill Notation):
C19H17ClF3NO3
CAS Number:
Molecular Weight:
399.79
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$204.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N[C@@H](CC1=CC=C(C(F)(F)F)C=C1)CC(O)=O)CC2=CC=CC(Cl)=C2

InChI key

QOSIJVVNNGXEKE-INIZCTEOSA-N

Biochem/physiol Actions

An acetamidophenylbutanoate derivative with inverse agonist and antagonist activity against human, but not mouse or rat, FFA2 (GPR43)
CATPB is an acetamidophenylbutanoate derivative with inverse agonist and antagonist activity against human, but not mouse or rat, FFA2 (GPR43). CATPB inhibits both constitutive and propionate (C3) -stimulated hFFA2 GTPγS incorporation (IC50 ~35 and 320 nM, respectively; [C3] = EC80, ~125 nM), as well as C3-induced hFFA2 G159E activity (IC50 ~400 nM). CATPB is shown to inhibit C3-induced intracellular ERK phosphorylation in hFFA2-expressing cells (IC50 ~3 μM against 10 mM C3).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liliana Íñiguez-Gutiérrez et al.
International journal of immunopathology and pharmacology, 34, 2058738420958949-2058738420958949 (2020-12-30)
Neutrophils represent the first line of host cellular defense against various pathogens. The most recently described microbicidal mechanism of these cells is the release of neutrophil extracellular traps (NET). Currently, a wide range of chemical and biological stimuli are known

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service