Skip to Content
Merck
All Photos(1)

Key Documents

SML1680

Sigma-Aldrich

Ex26

≥98% (HPLC)

Synonym(s):

1-[[[5′-[[1-(4-Chloro-3-methylphenyl)ethyl]amino]-2′-fluoro-3,5-dimethyl[1,1′-biphenyl]-4-yl]carbonyl]amino]-cyclopropanecarboxylic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$166.00
25 MG
$658.00

$166.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$166.00
25 MG
$658.00

About This Item

Empirical Formula (Hill Notation):
C28H28ClFN2O3
CAS Number:
Molecular Weight:
494.98
UNSPSC Code:
12352200
NACRES:
NA.77

$166.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C(C)NC2=CC=C(F)C(C3=CC(C)=C(C(NC4(CC4)C(O)=O)=O)C(C)=C3)=C2)C=C1C

InChI key

YVHNBORUVLWCKF-UHFFFAOYSA-N

Biochem/physiol Actions

Ex26 is a peripherally-restricted biphenylcarboxamide derivative that acts as a highly potent and subtype-selective S1P1 antagonist (IC50 = 0.93 nM against S1PR1 and >3.1 μM against S1PR2/3/4/5). Ex26 significantly elevates CpG-A-stimulated interferon (IFN) production in plasmacytoid dendritic cell culture (10 μM) and prevents S1P1 agonist-induced S1P1 receptor internalization and polyubiquitinylation in HEK cells (0.1-1,000 nM). Ex26 induces lymphocyte sequestration (ED50 = 0.06 mg/kg, i.p.) in lymph nodes and pulmonary edema (2 hr post 30 mg/kg i.p.) in mice, leading to the inhibition of lymphocyte and thymocyte egress (0.1 mg/kg/hr continuously for 3 days after a loading dose of 3 mg/kg). Ex26 has been shown to ameliorate experimental autoimmune encephalomyelitis (EAE) in a mouse MS model and elevate CpG-A-stimulated serum IFN upregulation in mice in vivo (30 mg/kg/d, i.p.).
Ex26 is a peripherally-restricted biphenylcarboxamide derivative that acts as a highly potent and subtype-selective S1P1 antagonist.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service