Skip to Content
Merck
All Photos(1)

Key Documents

SML3223

Sigma-Aldrich

CAY10566

≥98% (HPLC)

Synonym(s):

2-(6-(4-(2-Chloro-5-fluorophenoxy)piperidin-1-yl)pyridazin-3-yl)-5-methyl-1,3,4-oxadiazole, Abbott compound 74, Abbott-28c, CAY 10566, CAY-10566, SCD1 inhibitor 28c

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$291.00
25 MG
$1,180.00

$291.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$291.00
25 MG
$1,180.00

About This Item

Empirical Formula (Hill Notation):
C18H17ClFN5O2
CAS Number:
Molecular Weight:
389.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$291.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

off-white to orange

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Fc1cc(c(cc1)Cl)OC2CCN(CC2)c3nnc(cc3)c4nnc([o]4)C

InChI

1S/C18H17ClFN5O2/c1-11-21-24-18(26-11)15-4-5-17(23-22-15)25-8-6-13(7-9-25)27-16-10-12(20)2-3-14(16)19/h2-5,10,13H,6-9H2,1H3

InChI key

WFOFPVXMPTVOTJ-UHFFFAOYSA-N

Biochem/physiol Actions

CAY10566 is a potent and selective stearoyl-CoA desaturase 1 (SCD1) inhibitor (IC50 = 26 nM/hSCD1 and 4.5 nM/mSCD1; IC50 >10 μM/75 receptors, transporters, ion channels). CAY10566 effectively blocks the conversion of saturated long-chain fatty acid-CoAs (LCFA-CoAs) to monounsaturated LCFA-CoAs in HepG2 cells (IC50 = 7.9 nM by C17:1/C17:0 ratio; IC50 = 6.8 nM by C16:1/C16:0 ratio) and exhibits good metabolic stability (by rat & mouse liver microsome assays) and oral availability (rat F = 59.2%; 5 mg/kg p.o.). CAY10566 is typically used in the range from 0.1-10 μM for cell treatment.
Orally available, potent and selective stearoyl-CoA desaturase 1 (SCD1) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service