Skip to Content
Merck
All Photos(1)

Key Documents

SML1372

Sigma-Aldrich

Nifetepimine

≥95% (HPLC)

Synonym(s):

1,2,3,4-Tetrahydro-6-methyl-4-(3-nitrophenyl)-2-oxo-5-pyrimidinecarboxylic acid ethyl ester, 5-Ethoxycarbonyl-6-methyl-4-(3-nitrophenyl)-3,4-dihydropyrimidin-2(1H)-one, 6-Methyl-4-(3-nitrophenyl)-2-oxo-1,2,3,4-tetrahydropyrimidine-5-carboxylic acid ethyl ester, Ethyl-4-(3-nitrophenyl)-6-methyl-2-oxo-1,2,3,4 tetrahydropyrimidine-5 carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H15N3O5
CAS Number:
Molecular Weight:
305.29
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(OCC)C1=C(C)NC(NC1C2=CC([N+]([O-])=O)=CC=C2)=O

InChI

1S/C14H15N3O5/c1-3-22-13(18)11-8(2)15-14(19)16-12(11)9-5-4-6-10(7-9)17(20)21/h4-7,12H,3H2,1-2H3,(H2,15,16,19)

InChI key

PPBNQKLDXXHPMH-UHFFFAOYSA-N

Biochem/physiol Actions

Nifetepimine is a dihydropyrimidone calcium channel blocker that down-modulates SERCA3 (sarco/endoplasmic reticulum Ca2+ -ATPase 3) pump, and protects of the CD4+ T cells from tumor-induced apoptosis. Nifetepimine induces apoptotic cell death in triple negative breast cancer (TNBC) cells by stimulation of cross-talk between ER stress and the MEK/ERK pathway.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service