Skip to Content
Merck
All Photos(1)

Key Documents

SML1368

Sigma-Aldrich

CLP257

≥98% (HPLC)

Synonym(s):

(5Z)-5-[(4-Fluoro-2-hydroxyphenyl)methylene]-2-(tetrahydro-1-(2H)-pyridazinyl)-4(5H)-thiazolone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$178.00
25 MG
$722.00

$178.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$178.00
25 MG
$722.00

About This Item

Empirical Formula (Hill Notation):
C14H14FN3O2S
CAS Number:
Molecular Weight:
307.34
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$178.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to light brown

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1/C(SC(N2CCCCN2)=N1)=C/C3=CC=C(F)C=C3O

InChI

1S/C14H14FN3O2S/c15-10-4-3-9(11(19)8-10)7-12-13(20)17-14(21-12)18-6-2-1-5-16-18/h3-4,7-8,16,19H,1-2,5-6H2/b12-7-

InChI key

SKCADXVKQRCWTR-GHXNOFRVSA-N

Application

CLP257 has been used as a K+- Cl- cotransporter 2 (KCC2) enhancer in human neuron culture.[1]

Biochem/physiol Actions

CLP257 is a selective K+-Cl cotransporter KCC2 activator that restored impaired Cl transport in neurons with reduced KCC2 activity. Apparently, CLP257 modulates plasmalemmal KCC2 protein turnover post-translationally.
CLP257 is a selective K+-Cl cotransporter KCC2 activator.
In streptozotocin treated rats, by activating a K+- Cl- cotransporter 2 (KCC2) in the arginine-vasopressin neuron, CLP257 mediates γ-aminobutyric acid (GABA)ergic excitation.[2] It is however inactive with KCC1, KCC3 and KCC4.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aaron J Barbour et al.
Neurobiology of disease, 141, 104878-104878 (2020-04-29)
Approximately half of people infected with HIV (PWH) exhibit HIV-associated neuropathology (neuroHIV), even when receiving combined antiretroviral therapy. Opiate use is widespread in PWH and exacerbates neuroHIV. While neurons themselves are not infected, they incur sublethal damage and GABAergic disruption
Young-Beom Kim et al.
Diabetes, 67(3), 486-495 (2017-12-08)
Diabetes mellitus (DM) is associated with increased plasma levels of arginine-vasopressin (AVP), which may aggravate hyperglycemia and nephropathy. However, the mechanisms by which DM may cause the increased AVP levels are not known. Electrophysiological recordings in supraoptic nucleus (SON) slices
Martin Gagnon et al.
Nature medicine, 19(11), 1524-1528 (2013-10-08)
The K(+)-Cl(-) cotransporter KCC2 is responsible for maintaining low Cl(-) concentration in neurons of the central nervous system (CNS), which is essential for postsynaptic inhibition through GABA(A) and glycine receptors. Although no CNS disorders have been associated with KCC2 mutations

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service