Skip to Content
Merck
All Photos(1)

Key Documents

SML1308

Sigma-Aldrich

KT195

≥98% (HPLC)

Synonym(s):

[4-(4′-Methoxy[1,1′-biphenyl]-4-yl)-1H-1,2,3-triazol-1-yl](2-phenyl-1-piperidinyl)-methanone

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$513.00

$124.00


Check Cart for Availability


Select a Size

Change View
5 MG
$124.00
25 MG
$513.00

About This Item

Empirical Formula (Hill Notation):
C27H26N4O2
CAS Number:
Molecular Weight:
438.52
UNSPSC Code:
12352200
NACRES:
NA.77

$124.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[n]3(nnc(c3)c4ccc(cc4)c5ccc(cc5)OC)C(=O)N1C(CCCC1)c2ccccc2

InChI

1S/C27H26N4O2/c1-33-24-16-14-21(15-17-24)20-10-12-22(13-11-20)25-19-31(29-28-25)27(32)30-18-6-5-9-26(30)23-7-3-2-4-8-23/h2-4,7-8,10-17,19,26H,5-6,9,18H2,1H3

InChI key

OEHLNQDRVMDXQD-UHFFFAOYSA-N

Biochem/physiol Actions

KT195 is a potent (IC50 = 10 nM) and selective inhibitor of α/β-hydrolase domain-containing 6 (ABHD6), a serine hydrolase that acts as an alternative hydrolase of the endocannabinoid 2-arachidonoylglycerol (2-AG). KT195 has negligible activity against other serine hydrolases such as DAGLβ and has been used as a control probe for studies of DAGLβ as well as being a probe on its own to study ABHD6. KT195 treatment of Neuro2A cells caused significant accumulation of 2-AG. KT195 also lowered interleukin-1β (IL-1β) secretion from lipopolysaccharide-treated macrophages suggesting ABHD6 involvement in this activity as well.
KT195 is a potent (IC50 = 10 nM) and selective inhibitor of α/β-hydrolase domain-containing 6 (ABHD6).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service