Skip to Content
Merck
All Photos(2)

Key Documents

900883

Sigma-Aldrich

NIR-II dye

Synonym(s):

Near IR fluorescent dye

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$2,180.00

$2,180.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$2,180.00

About This Item

UNSPSC Code:
41116100
NACRES:
NA.23

$2,180.00


Check Cart for Availability

Request a Bulk Order

description

Mass extinction co-efficient: 3.5 Lg-1cm-1
viscous solid or liquid

Quality Level

form

liquid

λmax

780-810 nm

fluorescence

λem ~1050 nm, quantum yield ~2% (NIR emission)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

  • Stable solution in PBS. Miscible and stable in common buffers, serum and blood.
  • Store in dark at 4 °C.
  • Suitable excitation wavelengths: 785 nm, 808 nm, Emission: 900-1400 nm, peak ~1050 nm.
  • Quantum yield (QY) ~2% based on 0.5% QY of IR26 in 1,2-dichloroethane under 808 nm excitation. Quantum yield is ~4X of common Hipco single walled carbon nanotubes.
  • For in vivo imaging (e.g., vessels), inject ~100-150 μg in ~200 μL solution for a typical nude mouse but the dose can be varied.
  • Circulation half time of the dye is about 1 hour.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service