Skip to Content
Merck
All Photos(1)

Key Documents

SML1298

Sigma-Aldrich

ML309 hydrochloride

≥98% (HPLC)

Synonym(s):

N-[2-(Cyclopentylamino)-1-(2-methylphenyl)-2-oxoethyl]-N-(3-fluorophenyl)-1H-benzimidazole-1-acetamide hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$124.00
25 MG
$496.00

$124.00


Check Cart for Availability


Select a Size

Change View
5 MG
$124.00
25 MG
$496.00

About This Item

Empirical Formula (Hill Notation):
C29H29FN4O2 · HCl
CAS Number:
Molecular Weight:
521.03
UNSPSC Code:
12352200
NACRES:
NA.77

$124.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

ML309 is a potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (IDH1) (R132H and R132C) that binds reversibly the enzyme. ML309 inhibits production of neomorphic 2-hydroxyglutarate in glioblastoma cell line.
ML309 is a potent and selective inhibitor of mutant isocitrate dehydrogenase 1 (IDH1) (R132H and R132C).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Mauer et al.
Nature chemical biology, 15(4), 340-347 (2019-02-20)
Small nuclear RNAs (snRNAs) are core spliceosome components and mediate pre-mRNA splicing. Here we show that snRNAs contain a regulated and reversible nucleotide modification causing them to exist as two different methyl isoforms, m1 and m2, reflecting the methylation state

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service