Skip to Content
Merck
All Photos(1)

Key Documents

SML2498

Sigma-Aldrich

Vatalanib dihydrochloride

≥98% (HPLC)

Synonym(s):

1-(4-Chloroanilino)-4-(4-pyridylmethyl)phthalazine dihydrochloride, CGP 79787 dihydrochloride, CGP 79787D dihydrochloride, N-(4-Chloroanilino)-4-(pyridin-4-ylmethyl)phthalazin-1-amine dihydrochloride, PTK 787 dihydrochloride, PTK787 dihydrochloride, ZK 222584 dihydrochloride, ZK-222584 dihydrochloride, ZK222584 dihydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$116.00
50 MG
$391.00

$116.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$116.00
50 MG
$391.00

About This Item

Empirical Formula (Hill Notation):
C20H15ClN4 · 2HCl
CAS Number:
Molecular Weight:
419.73
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$116.00


Estimated to ship on13 April 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC(C=C1)=CC=C1NC2=NN=C(CC3=CC=NC=C3)C4=CC=CC=C42

InChI

1S/C20H15ClN4.2ClH/c21-15-5-7-16(8-6-15)23-20-18-4-2-1-3-17(18)19(24-25-20)13-14-9-11-22-12-10-14;;/h1-12H,13H2,(H,23,25);2*1H

InChI key

AZUQEHCMDUSRLH-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active anti-angiogenic agent that targets receptor tyrosine kinases VEGFR1/2/3 (FLT-1/KDR/FLT-4), PDGFRβ, c-KIT and c-FMS.
Vatalanib (PTK787, ZK 222584, CGP 79787D, CGP 79787 succinate) is an orally active anilinophthalazine derivative that exerts its anti-angiogenic efficacy by targeting receptor tyrosine kinase (RTK) VEGFR (VEGFR1/2 IC50 = 54/39 nM; IC50 ∼30 nM against 100 ng/mL VEGF-C-induced VEGFR3 autophosphorylation in MEVCs) and, to a less extent, PDGFRβ, c-kit, c-fms (IC50 = 364, 600, 567 nM, respectively) in an ATP-competitive manner with much reduced or no potency toward >90 other kinases. Typical dosing ranges: 0.1-10 μM in cultures, 5 nM-200 μM (zebrafish) and 30-100 mg/kg (mice & rats via po. or ip) in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service