Skip to Content
Merck
All Photos(1)

Key Documents

SML1284

Sigma-Aldrich

Efavirenz Ready Made Solution

10 mg/mL in DMSO

Synonym(s):

(4S)-6-Chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-2,4-dihydro-1H-3,1-benzoxazin-2-one, (4S)-6-Chloro-4-(2-cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$264.00

$264.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 ML
$264.00

About This Item

Empirical Formula (Hill Notation):
C14H9ClF3NO2
CAS Number:
Molecular Weight:
315.67
UNSPSC Code:
12352200
NACRES:
NA.77

$264.00


Check Cart for Availability

Request a Bulk Order

form

DMSO solution

Quality Level

concentration

10 mg/mL in DMSO

shipped in

dry ice

storage temp.

−20°C

SMILES string

FC(F)(F)[C@]2(OC(=O)Nc3c2cc(cc3)Cl)C#CC1CC1

InChI

1S/C14H9ClF3NO2/c15-9-3-4-11-10(7-9)13(14(16,17)18,21-12(20)19-11)6-5-8-1-2-8/h3-4,7-8H,1-2H2,(H,19,20)/t13-/m0/s1

InChI key

XPOQHMRABVBWPR-ZDUSSCGKSA-N

Biochem/physiol Actions

Efavirenz is a nonnucleoside reverse transcriptase inhibitor (NNRTI).
Efavirenz is a nonnucleoside reverse transcriptase inhibitor (NNRTI). Efavirenz is an anti-HIV drug, commonly used in combination therapy for AIDs treatment. Efavirenz is part of highly active antiretroviral therapy (HAART) for the treatment of a human immunodeficiency virus (HIV) type 1.
The plasma levels of this drug can determine the failure of treatment and CNS (central nervous system) side-effects in patients infected with HIV (human immunodeficiency virus). [1]

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Marzolini et al.
AIDS (London, England), 15(1), 71-75 (2001-02-24)
Limited information exists on the clinical usefulness of drug level monitoring for efavirenz, a once-daily non-nucleoside reverse transcriptase inhibitor (NNRTI). The aim of this study was to determine whether efavirenz plasma concentration monitoring could predict treatment failure and central nervous

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service