Skip to Content
Merck
All Photos(1)

Key Documents

5.05052

Sigma-Aldrich

Ezh2/Ezh1 Inhibitor, UNC1999

Synonym(s):

Ezh2/Ezh1 Inhibitor, UNC1999, Enhancer of Zested Homolog 1 Inhibitor, 1-Isopropyl-6-(6-(4-isopropylpiperazin-1-yl)pyridin-3-yl)-N-((6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl)-1H-indazole-4-carboxamide, Ezh1 Inhibitor, Ezh2 Inhibitor IV, HMTase Inhibitor XII, Histone Lysi, Enhancer of Zested Homolog 1 Inhibitor, 1-Isopropyl-6-(6-(4-isopropylpiperazin-1-yl)pyridin-3-yl)-N-((6-methyl-2-oxo-4-propyl-1,2-dihydropyridin-3-yl)methyl)-1H-indazole-4-carboxamide, Ezh1 Inhibitor, Ezh2 Inhibitor IV, HMTase Inhibitor XII, Histone Lysin

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$215.00

$215.00


Estimated to ship on29 May 2025

New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$215.00

About This Item

Empirical Formula (Hill Notation):
C33H43N7O2
CAS Number:
Molecular Weight:
569.74
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$215.00


Estimated to ship on29 May 2025

New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

CCCC1=C(C(=O)NC(=C1)C)CNC(=O)C2=C3C=NN(C3=CC(=C2)C4=CN=C(C=C4)N5CCN(CC5)C(C)C)C(C)C

General description

A cell-permeable GSK126 structural analog that acts as a potent, SAM-, but not substrate-, competitive inhibitor against Ezh1 and Ezh2 (IC50 = 5, 20, 20, 45 nM against PRC2 complex containing Ezh2 wt, Ezh2 Y641F, Ezh2 Y641N, or wt Ezh1 in H3 peptide methylation assays; [SAM] = 5 µM), while exhibiting much reduced or little potency toward panels of 15 other methyltransferases (IC50 >10 µM), 50 kinases (≤20% inhibition at 10µM), as well as 43 GPCRs, transporters, and ion channels, although high affinity binding is observed with Sigma-2 receptor S2R/PGRMC1 (Ki = 65 nM) with unknown impact toward its function. Cellular H3K27me3 level is shown to be largely reduced in MCF10A, MCF7, and DB cells with no significant effect on proliferation upon a 3-day UNC1999 treatment (ICmax = 3 to 5 µM), while complete cell death is reported in the EZH2 Y641N-harboring DLBCL line DB cell culture upon an 8-day 5 µM drug exposure. Exhibits good bioavailability in mice with the plasma drug concentration maintained above 100 nM for 20 h and 24 h, respectively, following a single 50 mg/kg oral or i.p. dosage (Cmax = 4.7 and >9.7 µM, respectively). Both its oral bioavailability and enhanced Ezh1 potency can greatly complement the Ezh2-selective GSK126 in in vitro and in vivo studies.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Ezh2
Reversible: yes
Secondary Target
Ezh1

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Konze, K.D., et al. 2013. ACS Chem. Biol.8, 1324.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service