Skip to Content
Merck
All Photos(1)

Key Documents

SML1283

Sigma-Aldrich

Cilastatin sodium Ready Made Solution

50 mg/mL in DMSO/H2O, 1:1

Synonym(s):

((Z)-7-[(2R)-2-Amino-3-hydroxy-3-oxopropyl]sulfanyl-2-{[(1S)-2,2-dimethylcyclopropanecarbonyl]amino}hept-2-enoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

2 ML
$311.00

$311.00


Check Cart for Availability


Select a Size

Change View
2 ML
$311.00

About This Item

Empirical Formula (Hill Notation):
C16H26N2O5S
CAS Number:
Molecular Weight:
358.45
UNSPSC Code:
12352200
NACRES:
NA.77

$311.00


Check Cart for Availability

form

liquid (1:1 DMSO:H2O)

Quality Level

concentration

50 mg/mL in DMSO/H2O, 1:1

shipped in

dry ice

storage temp.

−20°C

SMILES string

[Na+].S(C[C@@H](N)C(=O)[O-])CCCC\C=C(/NC(=O)[C@@H]1C(C1)(C)C)\C(=O)O

InChI

1S/C16H26N2O5S.Na/c1-16(2)8-10(16)13(19)18-12(15(22)23)6-4-3-5-7-24-9-11(17)14(20)21;/h6,10-11H,3-5,7-9,17H2,1-2H3,(H,18,19)(H,20,21)(H,22,23);/q;+1/p-1/b12-6-;/t10-,11-;/m1./s1

InChI key

QXPBTTUOVWMPJN-VQAYNFQASA-M

Biochem/physiol Actions

Cilastatin sodium is an inhibitor of the renal enzyme dehydropeptidase I and some bacterial zinc-containing β-lactamases.
Cilastatin sodium is an inhibitor of the renal enzyme dehydropeptidase I and some bacterial zinc-containing β-lactamases. By this action, cilastatin suppresses both host and target metabolism of the broad-spectrum antibiotic imipenem, improving its efficacy.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service