Skip to Content
Merck
All Photos(1)

Key Documents

SML1082

Sigma-Aldrich

Lamotrigine isethionate

≥98% (HPLC)

Synonym(s):

6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine mono(2-hydroxyethanesulfonate)

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$200.00
50 MG
$786.00

$200.00


Check Cart for Availability


Select a Size

Change View
10 MG
$200.00
50 MG
$786.00

About This Item

Empirical Formula (Hill Notation):
C9H7Cl2N5 · C2H6O4S
CAS Number:
Molecular Weight:
382.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$200.00


Check Cart for Availability

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

NC(N=N1)=NC(N)=C1C2=CC=CC(Cl)=C2Cl.OCCS(O)(=O)=O

InChI

1S/C9H7Cl2N5.C2H6O4S/c10-5-3-1-2-4(6(5)11)7-8(12)14-9(13)16-15-7;3-1-2-7(4,5)6/h1-3H,(H4,12,13,14,16);3H,1-2H2,(H,4,5,6)

InChI key

CJIDZLNMKONKAD-UHFFFAOYSA-N

Biochem/physiol Actions

Lamotrigine isethionate is an anticonvulsant, water soluble salt of Lamotrigine (Catalog No. L3791).
Lamotrigine isethionate is an anticonvulsant.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service