Skip to Content
Merck
All Photos(1)

Key Documents

SML3059

Sigma-Aldrich

Lacosamide

≥98% (HPLC)

Synonym(s):

(2R)-2-(Acetylamino)-N-benzyl-3-methoxypropanamide, ADD 243037, ADD-243037, ADD243037, Erlosamide, LCM, SPM-927, SPM927

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$103.00
25 MG
$417.00

$103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$103.00
25 MG
$417.00

About This Item

Empirical Formula (Hill Notation):
C13H18N2O3
CAS Number:
Molecular Weight:
250.29
UNSPSC Code:
12352200
NACRES:
NA.77

$103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 13 to 18° (C=0.5g/100mL, MeOH)

drug control

USDEA Schedule V

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N([C@H](COC)C(=O)NCc1ccccc1)C(=O)C

InChI

1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1

InChI key

VPPJLAIAVCUEMN-GFCCVEGCSA-N

Biochem/physiol Actions

Lacosamide is an orally active anticonvulsant that blocks voltage-gated sodium channels (VGSCs) selectively in their inactivated state (IC50 = 16 μM/Nav1.8, 182 μM/Nav1.7, 415 μM/Nav1.3) over resting state by enhancing slow inactivation. In addition to in vivo anticonvulsant activity (ED50 = 4.5 mg/kg in mice via i.p., 3.9 mg/kg in rats via p.o. by maximal electroshock seizure test), lacosamide also shows therapeutic efficacy in various pain models (10-30 mg/kg in rats via i.p.).
Orally active anticonvulsant that blocks voltage-gated sodium channels (VGSCs) selectively in inactivated over resting state by enhancing slow inactivation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service