Skip to Content
Merck
All Photos(1)

Key Documents

SML1022

Sigma-Aldrich

AS1949490

≥98% (HPLC)

Synonym(s):

3-[(4-Chlorophenyl)methoxy]-N-[(1S)-1-phenylethyl]- 2-thiophenecarboxamide, AS 1949490

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$114.00
25 MG
$449.00

$114.00


Check Cart for Availability
New, lower price on this item!


Select a Size

Change View
5 MG
$114.00
25 MG
$449.00

About This Item

Empirical Formula (Hill Notation):
C20H18ClNO2S
CAS Number:
Molecular Weight:
371.88
UNSPSC Code:
12352200
NACRES:
NA.77

$114.00


Check Cart for Availability
New, lower price on this item!

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(c(cc1)OCc3ccc(cc3)Cl)C(=O)N[C@@H](C)c2ccccc2

InChI

1S/C20H18ClNO2S/c1-14(16-5-3-2-4-6-16)22-20(23)19-18(11-12-25-19)24-13-15-7-9-17(21)10-8-15/h2-12,14H,13H2,1H3,(H,22,23)/t14-/m0/s1

InChI key

RFZPGNRLOKVZJY-AWEZNQCLSA-N

Biochem/physiol Actions

AS1949490 is a potent, selective SHIP2 phosphatase inhibitor.
AS1949490 is a potent, selective SHIP2 phosphatase inhibitor. AS1949490 activates insulin signaling pathways in liver and lowers glucose levels in diabetic mice.
AS1949490 is also known as (3-[(4-chlorobenzyl)oxy]-N-[(1S)-1-phenylethyl]-2-thiophenecarboxamide).[1] It enhances the phosphorylation of Akt, glucose consumption and glucose uptake in L6 myotubes. AS1949490 has the ability to block gluconeogenesis in FAO (cell line) hepatocytes.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Akira Suwa et al.
European journal of pharmacology, 642(1-3), 177-182 (2010-06-19)
Lipid phosphatase SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) plays an important role in the regulation of insulin signaling. In this report, we identified AS1938909, a novel small-molecule SHIP2 inhibitor. AS1938909 showed potent inhibition of SHIP2 (Ki=0.44 microuM) and significant selectivity
A Suwa et al.
British journal of pharmacology, 158(3), 879-887 (2009-08-22)
The lipid phosphatase known as SH2 domain-containing inositol 5'-phosphatase 2 (SHIP2) plays an important role in the regulation of the intracellular insulin signalling pathway. Recent studies have suggested that inhibition of SHIP2 could produce significant benefits in treatment of type
Gilbert Salloum et al.
Journal of cell science, 132(16) (2019-08-15)
Macropinocytosis is an actin-dependent but clathrin-independent endocytic process by which cells nonselectively take up large aliquots of extracellular material. Macropinocytosis is used for immune surveillance by dendritic cells, as a route of infection by viruses and protozoa, and as a
Abdelhalim Azzi
FEBS open bio, 10(10), 2191-2205 (2020-09-04)
Phosphatidylinositol (3,4,5)-trisphosphate (PI(3,4,5)P3) is required for protein kinase B (AKT) activation. The level of PI(3,4,5)P3 is constantly regulated through balanced synthesis by phosphoinositide 3-kinase (PI3K) and degradation by phosphoinositide phosphatases phosphatase and tensin homologue (PTEN) and SH2-domain containing phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service