Skip to Content
Merck
All Photos(1)

Key Documents

SML1806

Sigma-Aldrich

YU142670

≥98% (HPLC)

Synonym(s):

3-(4-pyridinyl)-1,2,4-Triazolo[3,4-b][1,3,4]thiadiazole, 3-(Pyridin-4-yl)-[1,2,4]triazolo[3,4-b][1,3,4]thiadiazole

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$221.00
25 MG
$878.00

$221.00


Check Cart for Availability


Select a Size

Change View
5 MG
$221.00
25 MG
$878.00

About This Item

Empirical Formula (Hill Notation):
C8H5N5S
CAS Number:
Molecular Weight:
203.22
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$221.00


Check Cart for Availability

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C12=NN=C(C3=CC=NC=C3)N1N=CS2

InChI key

HRBYAJQMVADJHW-UHFFFAOYSA-N

Biochem/physiol Actions

A selective OCRL1/INPP5F and OCRL2/INPP5B inhibitor that causes cellular PI(4,5)P2 accumulation and autophagosome-lysosome fusion defect.
YU142670 is a selective inhibitor against OCRL1/INPP5F (IC50 = 0.71 μM; substrate = PI(4,5)P2) and OCRL2/INPP5B (IC50 = 0.53 and 1.78 μM; substrate = PI3 and INPP5B/PI(4,5)P2, respectively) by targeting OCRL catalytic domain without affecting INPP5A, INPP5E, PTEN, SHP1, shrimp alkaline phosphatase, sphingomyelinase, or SYNJ1. YU142670 causes an increased PI(4,5)P2/PI4P ratio in human skin fibroblasts (by 50%; 50 μM for 1 h) and induces upregulated actin nucleation and ruffle activity at the plasma membrane without obvious cytotoxicity. Consistent with the negative regulatory role of PtdIns(4,5)P2 against the calcium channel mucolipin-1 (MCOLN1) that controls autophagosome-lysosome fusion, ehnahced autophagosome accumulation is observed in human kidney proximal tubule cells (PTCs) upon YU142670 (25 μM for 3 hr) or OCRL shRNA treatment.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Giovanna De Leo et al.
Nature cell biology, 18(8), 839-850 (2016-07-12)
Phosphoinositides (PtdIns) control fundamental cell processes, and inherited defects of PtdIns kinases or phosphatases cause severe human diseases, including Lowe syndrome due to mutations in OCRL, which encodes a PtdIns(4,5)P2 5-phosphatase. Here we unveil a lysosomal response to the arrival
Michelle Pirruccello et al.
ACS chemical biology, 9(6), 1359-1368 (2014-04-20)
Phosphoinositides are low abundance membrane phospholipids that have key roles in signaling, membrane trafficking, and cytoskeletal dynamics in all cells. Until recently, strategies for robust and quantitative development of pharmacological tools for manipulating phosphoinositide levels have focused selectively on PI(3,4,5)P3
Stephen J Mills et al.
Biochemistry, 55(9), 1384-1397 (2016-02-09)
The inositol polyphosphate 5-phosphatase INPP5B hydrolyzes the 5-phosphate group from water- and lipid-soluble signaling messengers. Two synthetic benzene and biphenyl polyphosphates (BzP/BiPhPs), simplified surrogates of inositol phosphates and phospholipid headgroups, were identified by thermodynamic studies as potent INPP5B ligands. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service