Skip to Content
Merck
All Photos(1)

Key Documents

SML0990

Sigma-Aldrich

JW651

≥98% (HPLC)

Synonym(s):

1,1,1,3,3,3-Hexafluoropropan-2-yl 4-(bis(4-chlorophenyl)methyl)piperazine-1-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$114.00
25 MG
$449.00

$114.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$114.00
25 MG
$449.00

About This Item

Empirical Formula (Hill Notation):
C21H18Cl2F6N2O2
CAS Number:
Molecular Weight:
515.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$114.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

ClC1=CC=C(C(C2=CC=C(Cl)C=C2)N3CCN(C(OC(C(F)(F)F)C(F)(F)F)=O)CC3)C=C1

InChI

1S/C21H18Cl2F6N2O2/c22-15-5-1-13(2-6-15)17(14-3-7-16(23)8-4-14)30-9-11-31(12-10-30)19(32)33-18(20(24,25)26)21(27,28)29/h1-8,17-18H,9-12H2

InChI key

UPIHQFKOCOKGEO-UHFFFAOYSA-N

Biochem/physiol Actions

JW651 is a potent selective inhibitor of MAGL, the enzyme predominantly responsible for the degradation of the endocannabinoid 2-arachidonoylglycerol (2-AG). JW651 inhibited mouse MAGL with an IC50 ~38 nM compared to 10,380 for ABHD6, a serine hydrolase that acts as an alternative hydrolase of 2-AG and >100,000 for FAAH, the hydrolase that degrades the endocannabinoid anandamide (AEA). JW651 was used as the MAGL inhibitor along with JW912, a fluorescent inihibitor of both MAGL and ABHD6.
JW651 is a potent selective inhibitor of MAGL; active in vivo.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Euna Yoo et al.
Cell chemical biology, 27(2), 143-157 (2020-01-25)
Salinipostin A (Sal A) is a potent antiplasmodial marine natural product with an undefined mechanism of action. Using a Sal A-derived activity-based probe, we identify its targets in the Plasmodium falciparum parasite. All of the identified proteins contain α/β serine

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

Related Content

The Cravatt group develops innovative technologies to understand enzyme roles in disease, focusing on activity-based protein profiling.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service