Skip to Content
Merck
All Photos(1)

Key Documents

SML0989

Sigma-Aldrich

H2L5186303

≥98% (HPLC)

Synonym(s):

(2Z,2′Z)-4,4′-[1,3-Phenylenebis(oxy-4,1-phenyleneimino)]bis[4-oxo-2-butenoic acid, 4,4′-[1,3-Phenylenebis(oxy-4,1-phenyleneimino)]bis[4-oxo-2-Butenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$122.00

$122.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$122.00

About This Item

Empirical Formula (Hill Notation):
C26H20N2O8
CAS Number:
Molecular Weight:
488.45
UNSPSC Code:
12352200
NACRES:
NA.77

$122.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

N(c1ccc(cc1)Oc2cc(ccc2)Oc3ccc(cc3)NC(=O)\C=C/C(=O)O)C(=O)\C=C/C(=O)O

InChI

1S/C26H20N2O8/c29-23(12-14-25(31)32)27-17-4-8-19(9-5-17)35-21-2-1-3-22(16-21)36-20-10-6-18(7-11-20)28-24(30)13-15-26(33)34/h1-16H,(H,27,29)(H,28,30)(H,31,32)(H,33,34)/b14-12-,15-13-

InChI key

HZFPOTBCYPWQSH-DZDAAMPGSA-N

Biochem/physiol Actions

H2L5186303 is specific antagonist of the lysophosphatidic acid receptor LPA2 (Ki = 7.2 nM). The compound H2L5186303 displays 40- to 1800-fold selectivity over other LPA receptors.
H2L5186303 is specific antagonist of the lysophosphatidic acid receptor LPA2.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service