Skip to Content
Merck
All Photos(1)

Key Documents

SML0743

Sigma-Aldrich

EUK-134

≥95% (HPLC)

Synonym(s):

(SP-5-13)-Chloro[[2,2′-[1,2-ethanediylbis[(nitrilo-?N)methylidyne]]bis[6-methoxyphenolato-KO]](2-)]-manganese, 2,2′-[1,2-Ethanediylbis(nitrilomethylidyne)]bis[6-methoxy-phenol manganese complex, Chloro[[2,2′-[1,2-ethanediylbis(nitrilomethylidyne)]bis[6-methoxyphenolato]](2-)-N2,N2′,O1,O1′]-manganese, EUK 134

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$109.00
50 MG
$283.00

$109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
$109.00
50 MG
$283.00

About This Item

Empirical Formula (Hill Notation):
C18H18ClMnN2O4
CAS Number:
Molecular Weight:
416.74
UNSPSC Code:
12352200
NACRES:
NA.77

$109.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

, faint to very dark brown

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[Mn+2]Cl.N(=Cc2c(c(ccc2)OC)[O-])CCN=Cc1c(c(ccc1)OC)[O-]

InChI

1S/C18H20N2O4.ClH.Mn/c1-23-15-7-3-5-13(17(15)21)11-19-9-10-20-12-14-6-4-8-16(24-2)18(14)22;;/h3-8,11-12,21-22H,9-10H2,1-2H3;1H;/q;;+3/p-3

InChI key

YUZJJFWCXJDFOQ-UHFFFAOYSA-K

Application

EUK-134 has been used as an antioxidant.[1]
EUK-134 has been used to determine the mechanism underlying hypoxia-inducible factor-1α (HIF-1α) stabilization in human aortic endothelial cells (HAECs).[1]

Biochem/physiol Actions

EUK-134 is a manganese-salen derivative that exhibit potent antioxidant activities and inhibits amyloid fibril formation.
EUK-134 is a manganese-salen derivative that exhibit potent antioxidant activities. EUK-134 is a superoxide dismutase (SOD) that inhibits amyloid fibril formation, including islet amyloid polypeptide (IAPP), β-amyloid and lysozyme amyloid aggregation. It appears that EUK-134 disrupts the pre-formed amyloid fibrils. EUK-134 increases the viability of the SK-N-MC cells exposed to preformed IAPP fibrils.

Other Notes

air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

HIF-1α is required for disturbed flow-induced metabolic reprogramming in human and porcine vascular endothelium.
Wu D, et al.
eLife, 6, e25217-e25217 (2017)
Anna K Stovall et al.
Frontiers in fungal biology, 2 (2021-10-09)
The fungal pathogen Cryptococcus neoformans relies on post-transcriptional mechanisms of gene regulation to adapt to stressors it encounters in the human host, such as oxidative stress and nutrient limitation. The kinase Gcn2 regulates translation in response to stress by phosphorylating
Cory L Simpson et al.
Cell reports, 34(5), 108689-108689 (2021-02-04)
The epidermis regenerates continually to maintain a protective barrier at the body's surface composed of differentiating keratinocytes. Maturation of this stratified tissue requires that keratinocytes undergo wholesale organelle degradation upon reaching the outermost tissue layers to form compacted, anucleate cells.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service