Skip to Content
Merck
All Photos(1)

Key Documents

341209

Sigma-Aldrich

EUK-8

A synthetic salen-manganese complex with high superoxide dismutase (SOD) and catalase-mimic activities, and oxyradical scavenging activities.

Synonym(s):

EUK-8

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$159.00

$159.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
$159.00

About This Item

Empirical Formula (Hill Notation):
C16H14ClMnN2O2
CAS Number:
Molecular Weight:
356.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$159.00


Estimated to ship on28 May 2025


Request a Bulk Order

Quality Level

Assay

≥90% (elemental analysis)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

dark brown

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[Mn+3].[Cl-].N(=C\c2c(cccc2)[O-])/CC\N=C\c1c(cccc1)[O-]

InChI

1S/C16H16N2O2.ClH.Mn/c19-15-7-3-1-5-13(15)11-17-9-10-18-12-14-6-2-4-8-16(14)20;;/h1-8,11-12,19-20H,9-10H2;1H;/q;;+3/p-3/b17-11+,18-12+;;

InChI key

HBQGFROGWOHBAG-SNTCFBDDSA-K

General description

A synthetic salen-manganese complex with high superoxide dismutase (SOD) and catalase-mimic activities, and oxyradical scavenging activities. Shown to have protective effects in numerous models for disease processes associated with oxidative stress, including inflammation, cardiovascular diseases, acute critical illness, autoimmune diseases, and neurological disorders. Useful in preventing cardiac damage caused by ischemic reperfusion. Also prevents β-amyloid peptide (Aβ)-induced free radical formation and lipid peroxidation. Imparts protection against Aβ40 and Aβ42 toxicity in hippocampal cultures.

Biochem/physiol Actions

Cell permeable: no
Primary Target
Superoxide dismutase (SOD) and catalase-mimic activities, and oxyradical scavenging activities
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 weeks at -20°C.

Other Notes

Colparet, E. E. and Lefebure, R. A. 2000. Br. J. Pharmacol.130, 359.
Brandier, C., et al. 1997. J. Vasc. Res. 34, 49.
Doctrow, S.R., et al. 1997. Adv. Pharmacol. 38, 247.
Bruce, A.J., et al. 1996. Proc. Natl. Acad. Sci. USA 93, 2312.
Pucheu, S., et al. 1996. Cardiovasc. Drug Ther. 10, 331.
Baudry, M., et al. 1993. Biochem. Biophys. Res. Commun. 192, 964.

Legal Information

Not available for sale in Africa, Australia, Great Britain, Europe, Latvia, New Zealand, and the United States.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service