Skip to Content
Merck
All Photos(1)

Documents

SML0378

Sigma-Aldrich

Ralfinamide mesylate

≥98% (HPLC)

Synonym(s):

(2S)-2-[[[4-[(2-Fluorophenyl)methoxy]phenyl]methyl]amino]-propanamide methanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19FN2O2 · CH3SO3H
CAS Number:
Molecular Weight:
398.45
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D 10 to 15° (c=1 HOAc)

color

white to beige

solubility

H2O: >15 mg/mL

storage temp.

2-8°C

SMILES string

CS(O)(=O)=O.C[C@H](NCc1ccc(OCc2ccccc2F)cc1)C(N)=O

InChI

1S/C17H19FN2O2.CH4O3S/c1-12(17(19)21)20-10-13-6-8-15(9-7-13)22-11-14-4-2-3-5-16(14)18;1-5(2,3)4/h2-9,12,20H,10-11H2,1H3,(H2,19,21);1H3,(H,2,3,4)/t12-;/m0./s1

InChI key

CHQVNINIGBRKGZ-YDALLXLXSA-N

Biochem/physiol Actions

Ralfinamide is a mixed sodium and N-type calcium channel blocker. Inhibiton is voltage-dependent, with higher potency against inactivated channels. Ralfinamide is two fold selective for tetrodotoxin (TTX)-resistant sodium channels compared with TTX-sensitive currents.

Features and Benefits

This compound is featured on the Calcium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Voltage-gated sodium channels are present in most excitable cell membranes and play an important role in generating action potentials.

We offers many products related to calcium channels for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service