Skip to Content
Merck
All Photos(1)

Key Documents

PZ0274

Sigma-Aldrich

PF-01247324

≥98% (HPLC)

Synonym(s):

6-Amino-5-(2,3,5-trichlorophenyl)pyridine-2-carboxylic acid methylamide, 6-Amino-N-methyl-5-(2,3,5-trichlorophenyl)-2-Pyridinecarboxamide, N-Methyl-6-amino-5-(2,3,5-trichlorophenyl)pyridine-2-carboxamide, PF 01247324, PF-1247324

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$127.00
25 MG
$510.00

$127.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Select a Size

Change View
5 MG
$127.00
25 MG
$510.00

About This Item

Empirical Formula (Hill Notation):
C13H10Cl3N3O
CAS Number:
Molecular Weight:
330.60
UNSPSC Code:
41121801
NACRES:
NA.77

$127.00


Check Cart for Availability
New, lower price on this item!

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

Clc1c(cc(cc1c2c(nc(cc2)C(=O)NC)N)Cl)Cl

InChI

1S/C13H10Cl3N3O/c1-18-13(20)10-3-2-7(12(17)19-10)8-4-6(14)5-9(15)11(8)16/h2-5H,1H3,(H2,17,19)(H,18,20)

InChI key

HPIUHDCRVYDAEJ-UHFFFAOYSA-N

Biochem/physiol Actions

In humans, PF-01247324 [6-amino-5-(2, 3, 5-trichloro-phenyl)-pyridine-2-carboxylic acid methylamide] prevents native tetrodotoxin-resistant (TTX-R) currents in dorsal root ganglion (DRG) neurons.[1]
PF-01247324 is an orally bioavailable, selective and potent NaV1.8 channel blocker that inhibits that exhibits analgesic efficacy in rodent behavioral models of pain.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

A novel selective and orally bioavailable Nav1. 8 channel blocker, PF?01247324, attenuates nociception and sensory neuron excitability.
Payne CE, et al.
British Journal of Pharmacology, 172(10), 2654-2670 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service