Skip to Content
Merck
All Photos(2)

Documents

SMB00298

Sigma-Aldrich

Fmoc-L-Asn((Ac)3-β-D-GlcNAc)-OH

≥95% (HPLC)

Synonym(s):

Fmoc-L-Asn((Ac)3-β-D-GlcNAc)-OH, Fmoc-L-Asn (β-D-GlcNAc(Ac)3)-OH, N-α-(fluorenylmethoxycarbonyl)-N-γ-(2-acetamido-3,4,6-tri-O-acetyl-2-deoxy-β-D-glucopyranosyl)-L-asparagine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H37N3O13
CAS Number:
Molecular Weight:
683.66
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (HPLC)

form

powder

functional group

Fmoc

storage temp.

−20°C

SMILES string

CC(O[C@@H]1[C@@H](NC(C)=O)[C@H](NC(C[C@H](NC(OCC2C3=C(C4=C2C=CC=C4)C=CC=C3)=O)C(O)=O)=O)O[C@H](COC(C)=O)[C@H]1OC(C)=O)=O

InChI

1S/C33H37N3O13/c1-16(37)34-28-30(48-19(4)40)29(47-18(3)39)26(15-45-17(2)38)49-31(28)36-27(41)13-25(32(42)43)35-33(44)46-14-24-22-11-7-5-9-20(22)21-10-6-8-12-23(21)24/h5-12,24-26,28-31H,13-15H2,1-4H3,(H,34,37)(H,35,44)(H,36,41)(H,42,43)/t25-,26+,28+,29+,30+,31+/m0/s1

InChI key

QOACISSHGAAMLK-MJCYOTSPSA-N

General description

A basic monosaccharide fragment of branched N-linked oligosaccharides found on glycoproteins.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service