Skip to Content
Merck
All Photos(1)

Key Documents

47516

Sigma-Aldrich

Fmoc-Cys(tBu)-OH

≥98.0% (HPLC)

Synonym(s):

Nα-Fmoc-S-tert-butyl-L-cysteine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$129.00

$129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$129.00

About This Item

Empirical Formula (Hill Notation):
C22H25NO4S
CAS Number:
Molecular Weight:
399.50
Beilstein:
4577031
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

$129.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (HPLC)

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)(C)SC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C22H25NO4S/c1-22(2,3)28-13-19(20(24)25)23-21(26)27-12-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h4-11,18-19H,12-13H2,1-3H3,(H,23,26)(H,24,25)/t19-/m0/s1

InChI key

IXAYZHCPEYTWHW-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

S N McCurdy
Peptide research, 2(1), 147-152 (1989-01-01)
Fmoc-Cys(t-Bu)-OH, Fmoc-Cys(Acm)-OH, and Fmoc-Cys(Trt)-OH exhibit excellent synthesis characteristics when used in Fmoc solid phase peptide synthesis on the Applied Biosystems Model 431A peptide synthesizer. The actual 5% scavenger mixture will vary according to the particular amino acid residues present. As

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service