Skip to Content
Merck
All Photos(1)

Key Documents

SMB00143

Sigma-Aldrich

Khelloside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$526.00

$526.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
$526.00

About This Item

Empirical Formula (Hill Notation):
C19H20O10
CAS Number:
Molecular Weight:
408.36
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

$526.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

COc1c2C(=O)C=C(CO[C@@H]3O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)Oc2cc4occc14

InChI

1S/C19H20O10/c1-25-18-9-2-3-26-11(9)5-12-14(18)10(21)4-8(28-12)7-27-19-17(24)16(23)15(22)13(6-20)29-19/h2-5,13,15-17,19-20,22-24H,6-7H2,1H3/t13-,15-,16+,17-,19-/m1/s1

InChI key

SJRACCTZSAUMGO-WIMVFMHDSA-N

General description

Natural product derived from plant source.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Stevens et al.
Arzneimittel-Forschung, 35(8), 1257-1260 (1985-01-01)
Khellin and khelloside (khellol glucoside) were examined in female cynomolgus monkeys to substantiate their ability to favorably modify serum lipoprotein cholesterol. Clinical chemistry parameters were also measured to obtain information indicative of possible drug toxicity. Both drugs were evaluated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service