Skip to Content
Merck
All Photos(1)

Key Documents

83580

Sigma-Aldrich

Reserpine

crystallized, ≥99.0% (HPLC)

Synonym(s):

(3β, 16β, 17α, 18β, 20α)-11,17-Dimethoxy-18-[(3,4,5-trimethoxybenzoyl)oxy]yohimban-16-carboxylic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$53.80
5 G
$177.00

$53.80


Available to ship on07 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 G
$53.80
5 G
$177.00

About This Item

Empirical Formula (Hill Notation):
C33H40N2O9
CAS Number:
Molecular Weight:
608.68
Beilstein:
102014
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$53.80


Available to ship on07 April 2025Details


Request a Bulk Order

Assay

≥99.0% (HPLC)

form

solid

optical activity

[α]20/D −123±3°, c = 1% in chloroform

quality

crystallized

mp

~265 °C (dec.)

SMILES string

CO[C@H]1[C@@H](C[C@@H]2CN3CCc4c([nH]c5cc(OC)ccc45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)c6cc(OC)c(OC)c(OC)c6

InChI

1S/C33H40N2O9/c1-38-19-7-8-20-21-9-10-35-16-18-13-27(44-32(36)17-11-25(39-2)30(41-4)26(12-17)40-3)31(42-5)28(33(37)43-6)22(18)15-24(35)29(21)34-23(20)14-19/h7-8,11-12,14,18,22,24,27-28,31,34H,9-10,13,15-16H2,1-6H3/t18-,22+,24-,27-,28+,31+/m1/s1

InChI key

QEVHRUUCFGRFIF-MDEJGZGSSA-N

Gene Information

human ... SLC18A2(6571)

Looking for similar products? Visit Product Comparison Guide

General description

Reserpine is an indole alkaloid that irreversibly binds vesicular monoamine transporters VMAT1 and VMAT2. It is a sympatholytic drug useful in treating systemic hypertension.[1]

Application

Reserpine has been used as a test chemical in performance studies.[2]

Biochem/physiol Actions

Inhibits vesicular uptake of catecholamines and serotonin.
Reserpine is used to treat hypertensive pregnancy difficulties.[1] This drug is also considered as antipsychotic and antihypertensive, to regulate high blood pressure.[3]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Repr. 1A - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

What All Should Know about Plant Drugs
Essentials of Biochemistry for Medical Students, 35-61 (2015)
Intra-laboratory validated human cell-based in vitro vasculogenesis/angiogenesis test with serum-free medium
Toimela T, et al.
Reproductive Toxicology, 70, 116-125 (2017)
Heart and circulatory system drugs and diuretics
Drugs During Pregnancy and Lactation, 193-236 (2007)
Julia Zwirchmayr et al.
Scientific reports, 10(1), 12323-12323 (2020-07-25)
In this study a robust, whole organism screening based on Caenorhabditis elegans is presented for the discovery of natural products (NP) with beneficial effects against obesity and age-related diseases. Several parameters of the elaborated workflow were optimized to be adapted
Zhen Lu et al.
The Journal of clinical investigation, 131(10) (2021-05-18)
Intercellular biomolecule transfer (ICBT) between malignant and benign cells is a major driver of tumor growth, resistance to anticancer therapies, and therapy-triggered metastatic disease. Here we characterized cholesterol 25-hydroxylase (CH25H) as a key genetic suppressor of ICBT between malignant and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service