SH1111
MISSION® shRNA Human Gene Family Set, Bacterial Glycerol Stock
Nuclear Hormone Receptors
About This Item
Recommended Products
product line
MISSION®
storage temp.
−70°C
Gene Information
human ... AR(367) , ESR1(2099) , ESR2(2100) , ESRRA(2101) , ESRRB(2103) , ESRRG(2104) , HNF4A(3172) , HNF4G(3174) , NR0B1(190) , NR0B2(8431) , NR1D1(9572) , NR1D2(9975) , NR1H2(7376) , NR1H3(10062) , NR1H4(9971) , NR1I2(8856) , NR1I3(9970) , NR2C1(7181) , NR2C2(7182) , NR2E1(7101) , NR2E3(10002) , NR2F2(7026) , NR2F6(2063) , NR3C2(4306) , NR4A1(3164) , NR4A2(4929) , NR4A3(8013) , NR5A1(2516) , NR5A2(2494) , NR6A1(2649) , PGR(5241) , PPARA(5465) , PPARD(5467) , PPARG(5468) , RARA(5914) , RARB(5915) , RARG(5916) , RORA(6095) , RORB(6096) , RORC(6097) , RXRA(6256) , RXRB(6257) , RXRG(6258) , THRA(7067) , THRB(7068) , VDR(7421)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Other Notes
Legal Information
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service