Skip to Content
Merck
All Photos(1)

Key Documents

SCP0017

Sigma-Aldrich

Amyloid Blocker

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$250.00

$250.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$250.00

About This Item

Empirical Formula (Hill Notation):
C42H63N9O8
Molecular Weight:
822.01
UNSPSC Code:
12352200
NACRES:
NA.32

$250.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Gln-Lys-Leu-Val-Phe-Phe-NH2

General description

Amyloid plaques characteristic of the Alzheimer brain contain fibrils composed of amyloid β aggregates. Peptides, small organic molecules and nanoparticles can be used to bind or aggregate amyloid β (Aβ). They have been used as a strategy for the development of pharmaceutical approaches or agents to treat Alzheimer′s disease (AD).[1] Short peptides containing the sequence KLVFF have been shown to bind specifically to the homologous region in Aβ and are used to prevent full length amyloid fibril formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of amyloid-? aggregation in Alzheimer's disease.
Wang Q
Current Pharmaceutical Design (2014)
L O Tjernberg et al.
The Journal of biological chemistry, 272(19), 12601-12605 (1997-05-09)
We have previously shown that short peptides incorporating the sequence KLVFF can bind to the approximately 40amino acid residue Alzheimer amyloid beta-peptide (Abeta) and disrupt amyloid fibril formation (Tjernberg, L. O., Näslund, J., Lindqvist, F., Johansson, J., Karlström, A. R.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service