Skip to Content
Merck
All Photos(3)

Documents

E6383

Sigma-Aldrich

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride

crystalline

Synonym(s):

EDC, N-Ethyl-N′-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDAC, EDC hydrochloride, WSC hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N3 · HCl
CAS Number:
Molecular Weight:
191.70
Beilstein:
5764110
EC Number:
MDL number:
UNSPSC Code:
12161703
PubChem Substance ID:
NACRES:
NA.28

product name

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline

form

crystalline

Quality Level

reaction suitability

reagent type: cross-linking reagent
reaction type: Peptide Synthesis

color

white to off-white

mp

110-115 °C (lit.)

solubility

H2O: ≤100 mg/mL

application(s)

advanced drug delivery
general analytical

storage temp.

−20°C

SMILES string

Cl.CCN=C=NCCCN(C)C

InChI

1S/C8H17N3.ClH/c1-4-9-8-10-6-5-7-11(2)3;/h4-7H2,1-3H3;1H

InChI key

FPQQSJJWHUJYPU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-(3-dimethylaminopropyl) carbodiimide hydrochloride, commonly known as EDAC HCl, is a potent and widely employed water-soluble reagent in chemical and biochemical research due to its pivotal role in facilitating amide bond formation. In peptide synthesis, EDC HCl demonstrates efficiency by coupling amino acids through their carboxyl and amine groups, thereby forming peptide backbones. This capability is particularly valuable for creating peptides with specific sequences and functionalities.

Beyond peptides, EDC HCl extends its influence to the construction of immunogens, where it covalently attaches haptens (small immune-response eliciting molecules) to carrier proteins, playing an instrumental role in vaccine research. The versatility of EDAC HCl further unfolds in its ability to modify nucleic acids, allowing for the labeling of DNA and RNA through their 5′ phosphate groups. This facilitates the visualization, tracking, and analysis of these essential molecules, contributing to advancements in nucleic acid research.

Additionally, EDAC HCl serves as a biomolecule bridge by acting as a crosslinker, connecting amine-reactive NHS-esters of biomolecules to carboxyl groups. This technique proves valuable in protein conjugation, enabling the creation of hybrid molecules with novel properties and functions. The underlying mechanism of EDAC HCl involves its reaction with a carboxyl group, forming an unstable intermediate that actively seeks an amine partner. The delicate balance of this reaction underscores the importance of optimizing conditions for efficient conjugation. The assistance of N-hydroxysuccinimide (NHS) further enhances EDAC HCl′s capabilities, stabilizing the intermediate and enabling two-step conjugation procedures. This additional feature provides greater flexibility and control, particularly in dealing with complex biomolecules.

Application

N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride has been used:
  • for the immobilisation of trypsin onto self-assembled monolayers (SAMs)
  • as a component for the preparation of collagen matrices
  • for the preparation of phosphoethanolamine(PEt)-conjugated sepharose

Biochem/physiol Actions

Water soluble condensing reagent. EDAC is generally utilized as a carboxyl activating agent for amide bonding with primary amines. In addition, it will react with phosphate groups. EDAC has been used in peptide synthesis; crosslinking proteins to nucleic acids; and preparation of immunoconjugates as examples. Typically, EDAC is utilized in the pH range 4.0-6.0 without buffers. In particular, amine and carboxylate buffers should be avoided.

Features and Benefits

Versatile and adaptable for a wide variety of laboratory and research applications

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Stomach,large intestine,lymph node

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrzej S Pitek et al.
Biomaterials, 89, 89-97 (2016-03-08)
Plant virus-based nanoparticles (VNPs) are a novel class of nanocarriers with unique potential for biomedical applications. VNPs have many advantageous properties such as ease of manufacture and high degree of quality control. Their biocompatibility and biodegradability make them an attractive
The phosphocholine-binding pocket on C-reactive protein is necessary for initial protection of mice against pneumococcal infection
Gang TB, et al.
The Journal of Biological Chemistry, jbc-M112 (2012)
Junhong Yan et al.
ACS sensors, 5(4), 1168-1176 (2020-03-20)
The ability to continuously measure concentrations of small molecules is important for biomedical, environmental, and industrial monitoring. However, because of their low molecular mass, it is difficult to quantify concentrations of such molecules, particularly at low concentrations. Here, we describe
Development and characterization of a collagen-based matrix for vascularization and cell delivery.
Ellis CE, et al
BioResearch Open Access, 4(1), 188-197 (2015)
Miaobo Ye et al.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 85, 153401-153401 (2020-11-17)
The Coronavirus disease 2019 pneumonia broke out in 2019 (COVID-19) and spread rapidly, which causes serious harm to the health of people and a huge economic burden around the world. In this study, the network pharmacology, molecular docking and surface

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service