Skip to Content
Merck
All Photos(1)

Key Documents

SAE0056

Sigma-Aldrich

Rapid Chemi AP Substrate

aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
$909.00
500 ML
$3,680.00
1000 ML
$6,370.00

$909.00


Check Cart for Availability


Select a Size

Change View
100 ML
$909.00
500 ML
$3,680.00
1000 ML
$6,370.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$909.00


Check Cart for Availability

Product Name

Rapid Chemi AP Substrate,

grade

Proteomics Grade

Quality Level

form

aqueous solution

storage temp.

2-8°C

General description

Rapid ChemiAP Substrate is a sensitive, chemiluminescent substrate for alkaline phosphatase that allows for the rapid, reproducible detection of alkaline phosphatase-labeled molecules

Application

Rapid ChemiAP Substrate is supplied as a ready-to-use aqueous solution, i.e., no dilution is necessary, for use in a variety of membrane-based applications such as Western Blotting and ELISA. Other applications include Southern, Northern Blotting, detection of placental alkaline phosphatase, detection of secreted alkaline phosphatase, reporter gene assays, nucleic acid probe assays and alkaline or acid phosphatase biomarker detection (stem cells and tumor markers).

Caution

Do not use diluents or wash buffers that contain EDTA or other chelators as they can remove divalent cations (Mg+2, Zn+2) and cause loss of AP activity.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service