Skip to Content
Merck
All Photos(1)

Key Documents

69086

Sigma-Aldrich

CDP-Star® AP Substrate

Synonym(s):

AP Substrate, CDP-Star Detection Reagent, CDP-Star Substrate

Sign Into View Organizational & Contract Pricing

Select a Size

40 ML
$511.00

$511.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
40 ML
$511.00

About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

$511.00


Estimated to ship on28 May 2025


Request a Bulk Order

form

liquid

Quality Level

manufacturer/tradename

Novagen®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

2-8°C

General description

The CDP-Star AP Substrate enables subnanogram sensitivity in a convenient ready-to-use format, and includes Nitro-Block II signal enhancer for increased signal-to-noise ratios with standard nitrocellulose membranes.

Warning

Toxicity: Irritant (B)

Legal Information

CDP-Star is a registered trademark of Tropix, Inc.
NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service