Skip to Content
Merck
All Photos(2)

Documents

SAB5900205

Sigma-Aldrich

Alpaca Anti-Her2, recombinant single domain Antibody

new

Recombinant Alpaca (VHH) Nano antibody

Synonym(s):

CD340, HER-2, HER2, c-ERB2, p185(erbB2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

alpaca

Quality Level

recombinant

expressed in E. coli

conjugate

unconjugated

antibody form

affinity purified VHH antibody

clone

EB02, monoclonal

Assay

>95% (SDS-PAGE)

form

lyophilized solid

mol wt

13.7 kDa
~15-18 kDa

purified by

immobilized metal affinity chromatography (IMAC)

species reactivity

human

concentration

1 mg/mL

technique(s)

immunofluorescence: 1:200-1:600
immunohistochemistry: 1:200-1:600

color

colorless

pH

( in PBS, pH 7.4)

isotype

VHH

Protein ID accession no.

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ERBB2(2064)

General description

Receptor tyrosine-protein kinase erbB-2, also known as Her2, is a protein that in humans is encoded by the ERBB2 gene. The ErbB family consists of four closely related tyrosine kinase receptors that act as potent mediators of normal cell growth and development. Aberrant expression or function of one or more of these receptors can play a major role in the development and evolution of cancer. Her2 has been implicated in the evolution of both breast and gastric cancers, and is evident in other cancer types such as ovarian and salivary gland tumors. Her2 possesses an active tyrosine kinase domain, but no direct ligand has been identified yet. Her2 is the preferred binding partner to the other members of the ErbB family and is thought to act primarily through the Ras-MAPK, PI3k-PKB/Akt, and PLC-PKC signalling pathways.Alpaca derived anti-HER2 VHH single domain antibody (molecular weight: 13.7 kDa), expressed in E. coli under conditions free from animal derived components. It has a 6His tag at its C-terminal.
Single-domain antibody (sdAb), also called nano-antibody, is the variable region isolated from the antigen-binding variable domain (VHH) of camelid heavy-chain-only antibody. It lacks light chains and a first constant domain(CH1). This configuration confers a number of unique advantages over conventional monoclonal antibodies. The small size of just 15 kDa (2~4 nm), single-domain antibody is the smallest antigen-binding fragment that can easily penetrate tissues, allowing them to bind epitopes that are inacessible to conventional antibodies. These single-domain antibodies offer superior chemical and thermal stability and increased solubility than conventional antibodies. All our antibodies are tested to ensure the highest level of specificity, sensitivity and reproducibility.

Specificity

Human Epidermal Growth Factor Receptor 2 (Her2)

Immunogen

Recombinant human HER2 (Thr23-Thr652) protein

Application

IHC,IF

Packaging

50 µg per vial / 100 µg per vial

Physical form

Affinity purified VHH single domain antibody lyophilized from PBS, pH7.8

Reconstitution

Reconstitute the lyophilized antibody with sterile deionized water or with 50% glycerol (ACS grade or better) to a stock solution of 1 mg/mL. Solubilize for 30 to 60 minutes at room temperature with occasional gentle mixing. Avoid vigorous shaking or vortexing.

Storage and Stability

Stable for 1 year at 4°C from date of receipt.

Other Notes

Storage at 4°C; Do not freeze.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service