Skip to Content
Merck
All Photos(2)

Key Documents

SAB5801049

Sigma-Aldrich

Alpaca Anti-Chicken IgY single domain Antibody

recombinant Alpaca(VHH) Nano antibody

Synonym(s):

Recombinant Alpaca single variable domain secondary Nano antibody

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$196.00

$196.00


Check Cart for Availability


Select a Size

Change View
100 μG
$196.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$196.00


Check Cart for Availability

biological source

alpaca

recombinant

expressed in E. coli

conjugate

unconjugated (6*his tag and one cys were added at the C terminal of the VHH)

antibody form

affinity purified immunoglobulin

antibody product type

secondary antibodies

clone

recombinant monoclonal

form

lyophilized solid

mol wt

14 kDa

species reactivity

Alpaca, chicken

technique(s)

ELISA: 1:10000-1:50000
western blot: 1:10000-1:50000

size

0.1 mL

color

colorless

isotype

VHH

shipped in

wet ice

storage temp.

2-8°C

General description

Alpaca Anti-Chicken IgY single domain Antibody is designed to specifically detect chicken IgY. It is a monovalent, single domain, recombinant antibody derived from the variable region of heavy chain of Alpaca pacous. Based on immunoelectrophoresis and/or ELISA, Alpaca Anti-Chicken IgY single domain Antibody detects the chicken IgY selectively but not with mouse, rabbit, human, cynomolgus, rat, goat IgG.
Hens egg yolk immunoglobulins IgY can be transferred from the serum of mother hen to the offspring egg yolk to acquire immunity. In the immunodiagnostic technologies, IgY is an excellent antibody for using in immunological assays involving mammalian sera, due to discriminative properties of IgY compared to mammalian IgG, as IgY does not react with the rheumatoid factor and human anti-mouse IgG antibodies do not activate the complement system and do not bind to Fc receptor. Also, they have poor cross reactivity to mammalian IgG due to immunological differences. IgY does not contain a hinge region but does contain an additional constant domain. The whole IgY molecule possesses both the Fc region and the Fab region, which possessing the epitope-recognition site. The IgYcontains two heavy and light chains. The common IgY is monomeric with a molecular weight of approximately 170 kDa.
VHH are single-domain antibodies derived from the variable regions of heavy chain of Camelidae immunoglobulin. The size of VHH is extremely small(less than15KDa) compared to other forms of antibody fragment, which significantly increase the permeability of VHH. Thus VHH is considered of great value for research, diagnostics and therapeutics.

Specificity

Fc region of chicken IgY
Isotype: VHH domain of alpaca IgG2b/2c

Immunogen

Recombinant Alpaca single variable domain secondary nano antibody

Packaging

100 μg in Glass bottle

Physical form

Lyophilized from 10mM PBS (pH 7.5), 0.05% sucrose, 0.1% trehalose

Reconstitution

Add 100uL dH2O or 100uL dH2O with 50% Glycerol to a final concentration of 1mg/mL

Storage and Stability

After reconstitution, store at –20 °C(Avoid freeze / thaw cycles). Stable for 12 months at -20°C

Other Notes

No cross-reactivity with mouse, rabbit, human, cynomolgus, rat, goat IgG

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service