Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702670

Sigma-Aldrich

Anti-HOOK2 Antibody, clone 8C9Q8, Rabbit Monoclonal

Synonym(s):

HOOK2, HK2, protein Hook homolog 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$641.00

$641.00


Estimated to ship on01 May 2025



Select a Size

Change View
100 μL
$641.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$641.00


Estimated to ship on01 May 2025


biological source

rabbit

Quality Level

material

colorless

clone

8C9Q8, monoclonal

form

liquid

mol wt

90 kDa

species reactivity

rat, rat, human

concentration

0.50 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MSVDKAELCGSLLTWLQTFHVPSPCASPQDLSSGLAVAYVLNQIDPSWFNEAWLQGISEDPGPNWKLKVSNLKMVLRSLVEYSQDVLAHPVSEEHLPDVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HOOK2(29911)

General description

Hook proteins are cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules, and more divergent C-terminal domains, which mediate binding to organelles. The Drosophila Hook protein is a component of the endocytic compartment.[supplied by OMIM, Apr 2004]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-100 of human HOOK2 (Q96ED9).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service