Skip to Content
Merck
All Photos(2)

Key Documents

MAB1618-AF647

Sigma-Aldrich

Anti-Dynein Antibody, IC, cytosolic, clone 74.1, Alexa Fluor 647 conjugate

clone 74.1, from mouse, ALEXA FLUOR 647

Synonym(s):

Cytoplasmic dynein 1 intermediate chain 1, Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1, Cytoplasmic dynein 1 intermediate chain 2, Cytoplasmic dynein intermediate chain 2, Dynein intermediate chain 2, cytosoli

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$489.00

$489.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$489.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$489.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 647

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

74.1, monoclonal

species reactivity

sheep, mouse, Xenopus, human, Drosophila, rat, bovine, fish

technique(s)

immunocytochemistry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... DYNC1I1(1780)

General description

Cytoplasmic dynein 1 intermediate chain 1 (UniProt Q29RQ3; also known as Cytoplasmic dynein intermediate chain 1, Dynein intermediate chain 1, cytosolic, DH IC-1) and Cytoplasmic dynein 1 intermediate chain 2 (UniProt Q0III3; also known as Cytoplasmic dynein intermediate chain 2, Dynein intermediate chain 2, cytosolic, DH IC-2) are encoded by the DYNC1I1 gene (Gene ID 613724) and DYNC1I2 gene (Gene ID 526329) in bovine, respectively. Cytoplasmic dynein complexes are microtubules-associated retrograde transport motors. Cytoplasmic dynein 1 is the more abundant motor in cells, while cytoplasmic dynein 2 takes part in intraflagellar transport. Cytoplasmic dynein 1 is a multisubunit complex of ∼1.5 MDa composed of a homodimer of heavy chains (encoded by DYNC1H1), two intermediate chains (encoded by DYNC1I1 and DYNC1I2), light-intermediate chains (encoded by DYNC1LI1, DYNC1LI2) and light chains (encoded by DYNLT1, DYNLT3, DYNLRB1, DYNLRB2, DYNLL1, DYNLLl2). The heavy chain homodimer acts as the dynein complex core and binds microtubules to enable ATP-dependent cytoplasmic dynein movement. The other dynein subunits associate as homodimers and play regulatory roles in maintaining the complex stability, modulating its activity, and mediating its interaction with accessory and cargo proteins. The intermediate chain proteins (IC1 & IC2) are involved in cargo binding and specificity, either alone or via interaction with the dynactin complex. IC1 and IC2 share 69% protein identity. They interact with the dynein light chains and the p150 subunit of dynactin at the N-terminus and with the heavy chains through WD40 repeats at the intermediate chain C-terminus.

Specificity

Reacts with all known forms of the Mr = 74,000 Dalton intermediate chain subunit (IC74) of mammaliam cytoplasmic dynein. The antigenic site has been localized to the first 60 amino acids of the amino terminal portion of the molecule (a region conserved in both IC74 genes).

Immunogen

Bovine brain cytoplasmic dynein.
Epitope: Within N-terminal 60 amino acids.

Application

Anti-Dynein Antibody, IC, cytosolic, clone 74.1, Alexa Fluor 647 conjugate is an antibody against Dynein for use in Immunocytochemistry.
Immunocytochemistry Analysis: An 1:100 dilution of this antibody from a representative lot detected Dynein in NIH/3T3 cells.
The unconjugated antibody (Cat. No. MAB1618) is shown to be suitable also for Western blotting, immunocytochemistry, immunofluorescence, and immunoprecipitation applications.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton

Quality

Evaluated by Immunocytochemistry in A431 cells.

Immunocytochemistry Analysis: An 1:100 dilution of this antibody detected Dynein in A431 cells.

Target description

~74 kDa observed.

Physical form

Protein A purified
Purified mouse monoclonal IgG2b antibody conjugate in PBS with 15 mg/mL BSA and 0.1 % sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service