Skip to Content
Merck
All Photos(1)

Key Documents

SAB5300210

Sigma-Aldrich

Monoclonal Anti-RICTOR antibody produced in mouse

clone 7B3, ascites fluid

Synonym(s):

DKFZp686B11164, KIAA1999, MGC39830, PIA, mAVO3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$642.00

$642.00


Check Cart for Availability


Select a Size

Change View
100 μL
$642.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$642.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

7B3, monoclonal

mol wt

192 kDa

species reactivity

monkey, mouse, human

technique(s)

direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
immunohistochemistry: 1:200-1:1,000
indirect immunofluorescence: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RICTOR(253260)

Immunogen

Purified recombinant fragment of human RICTOR expressed in E.coli.
Mouse monoclonal antibody raised against RICTOR

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lifeng Wang et al.
Oncology letters, 14(5), 6198-6202 (2017-11-09)
In order to understand the clinical significance of rapamycin-insensitive companion of mammalian target of rapamycin (Rictor) in colorectal cancer (CRC), 62 CRC tissue samples excised during operations were evaluated by immunohistochemistry. Analysis of the association between the expression level of
Yi Yang et al.
Chemical biology & drug design, 99(6), 884-896 (2022-03-22)
Alzheimer's disease (AD) is a chronic neurodegenerative disorder that can cause cognitive impairment. Ginsenoside Rg1 (Rg1) has a significant neuroprotective effect on animals with memory impairment. However, the mechanism of how Rg1 mediates the Wnt signaling pathway and improves cognitive
Rita Verma et al.
The Journal of biological chemistry, 290(11), 6810-6824 (2015-01-22)
G protein-coupled receptor (GPCR) signaling mediates many cellular functions, including cell survival, proliferation, and cell motility. Many of these processes are mediated by GPCR-promoted activation of Akt signaling by mammalian target of rapamycin complex 2 (mTORC2) and the phosphatidylinositol 3-kinase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service