Skip to Content
Merck
All Photos(1)

Key Documents

SAB5201889

Sigma-Aldrich

Monoclonal Anti-HSP70-Biotin antibody produced in mouse

clone 1H11

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$472.00

$472.00


Check Cart for Availability


Select a Size

Change View
100 μG
$472.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

$472.00


Check Cart for Availability

biological source

mouse

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1H11, monoclonal

form

buffered aqueous solution

mol wt

70 kDa

species reactivity

human

concentration

1.0 mg/mL

technique(s)

flow cytometry: 1:250
immunofluorescence: 1:100
western blot: 1:1000

isotype

IgG1

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA1A(3303)

Related Categories

Immunogen

Human native HSP70 protein

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service